Skip to Content
Merck
All Photos(1)

Key Documents

SML3501

Sigma-Aldrich

Dehydrocorydaline

≥98% (HPLC)

Synonym(s):

13-Methylpalmatine, 5,6-Dihydro-2,3,9,10-tetramethoxy-13-methyldibenzo[a,g]quinolizinium

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00
25 MG
$417.00

$103.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$103.00
25 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C22H24NO4
CAS Number:
Molecular Weight:
366.43
MDL number:
UNSPSC Code:
41115709
NACRES:
NA.77

$103.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=C2[N+](CCC3=C2C=C(OC)C(OC)=C3)=CC4=C(OC)C(OC)=CC=C41

InChI

1S/C22H24NO4/c1-13-15-6-7-18(24-2)22(27-5)17(15)12-23-9-8-14-10-19(25-3)20(26-4)11-16(14)21(13)23/h6-7,10-12H,8-9H2,1-5H3/q+1

InChI key

RFKQJTRWODZPHF-UHFFFAOYSA-N

Biochem/physiol Actions

Alkaloidal isolated from Rhizoma corydalis that exhibits anti-inflammatory, antinociceptive and anti-tumor effects
Dehydrocorydaline is an alkaloidal isolated from Rhizoma corydalis that exhibits anti-inflammatory, antinociceptive and anti-tumor effects. Dehydrocorydaline attenuates bone cancer pain through suppression M1 phenotype and upregulation M2 phenotype of microglia in the spinal cord.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydrocorydaline attenuates bone cancer pain by shifting microglial M1/M2 polarization toward the M2 phenotype.
Huo W, Zhang Y, Liu Y, Lei Y, Sun R, Zhang W, Huang Y, Mao Y, Wang C, Ma Z, Gu X.
Molecular Pain (2018)
Zhi-Yu Yin et al.
Scientific reports, 6, 27129-27129 (2016-06-09)
Dehydrocorydaline (DHC) is an alkaloidal component isolated from Rhizoma corydalis. Previous studies have shown that DHC has anti-inflammatory and anti-tumor effects and that it can protect the cardiovascular system. However, there are few studies of the antinociceptive effects of DHC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service