Skip to Content
Merck
All Photos(1)

Key Documents

SML3499

Sigma-Aldrich

LM10

≥98% (HPLC)

Synonym(s):

(E)-3-(2-(1H-tetrazol-5-yl)vinyl)-6-fluoro-1H-indole, 6-Fluoro-3-[(1E)-2-(2H-tetrazol-5-yl)ethenyl]-1H-indole, LM 10, trans-6-Fluoro-3-(2-(1H-tetrazol-5-yl)vinyl)-1H-indole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00
25 MG
$417.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$103.00
25 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C11H8FN5
CAS Number:
Molecular Weight:
229.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC2=C(C=C1)C(/C=C/C3=NN=NN3)=CN2

InChI

1S/C11H8FN5/c12-8-2-3-9-7(6-13-10(9)5-8)1-4-11-14-16-17-15-11/h1-6,13H,(H,14,15,16,17)/b4-1+

InChI key

JDBSZVDIUIRSDG-DAFODLJHSA-N

Biochem/physiol Actions

LM10 is an orally available, aqueous soluble, potent, selective and non-toxic tryptophan 2,3-dioxygenase (TDO) inhibitor. LM10 diminishes tryptophan degradation and effectively prevents the growth of TDO-expressing P815 tumor cells by reversing immune resistance in immune competent but not in immunodeficient mice (at a dose of 4 mg/day, 1 mg/ml in water, pH 9.0). LM10 suppresses autoimmune reactions induced by mouse hepatitis virus in mice.
Orally available, potent and selective tryptophan 2,3-dioxygenase (TDO) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luc Pilotte et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(7), 2497-2502 (2012-02-07)
Tryptophan catabolism mediated by indoleamine 2,3-dioxygenase (IDO1) is an important mechanism of peripheral immune tolerance contributing to tumoral immune resistance, and IDO1 inhibition is an active area of drug development. Tryptophan 2,3-dioxygenase (TDO) is an unrelated hepatic enzyme that also
Abdulla A-B Badawy
Medical hypotheses, 131, 109314-109314 (2019-08-25)
Metabolic targeting of liver 5-aminolevulinate synthase (5-ALAS) by inhibition of heme utilisation by tryptophan (Trp) 2,3-dioxygenase (TDO) or the use of tryptophan is proposed as a therapy of acute hepatic porphyrias. 5-ALAS, the rate-limiting enzyme of heme biosynthesis, is under
Maite Duhalde Vega et al.
Immunology letters, 217, 25-30 (2019-11-15)
In a previous work we demonstrated that inhibition of mouse indoleamine 2,3-dioxygenase (IDO) by methyltryptophan (MT) exacerbated the pathological actions of mouse hepatitis virus (MHV-A59) infection, suggesting that tryptophan (TRP) catabolism was involved in viral effects. Since there is a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service