Skip to Content
Merck
All Photos(1)

Key Documents

SML3465

Sigma-Aldrich

Toosendanin

≥90% (HPLC)

Synonym(s):

(1S,2aR,3aS,3bS,4R,5aR,6R,7S,9S,9aS,9bR,11R,11aR,14S)-1-(furan-3-yl)-4,9,14-trihydroxy-3b,6,11a-trimethyl-10-oxotetradecahydro-1H-6,9a-(methanooxymethano)naphtho[1′,2′:6,7]indeno[1,7a-b]oxirene-7,11-diyl diacetate, 28-Deacetylsendanin, Chuanliansu

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$103.00
50 MG
$417.00

$103.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$103.00
50 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C30H38O11
CAS Number:
Molecular Weight:
574.62
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

$103.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

C[C@@]1([C@H]2OC(C)=O)[C@]3([C@]([H])(O3)C[C@H]1C4=COC=C4)[C@]5(C)[C@H](O)C[C@@]6([H])[C@@](C)([C@@H]7O)[C@H](OC(C)=O)C[C@H](O)[C@]6(CO7)[C@@]5([H])C2=O

InChI

1S/C30H38O11/c1-13(31)39-20-10-19(34)29-12-38-25(36)26(20,3)17(29)9-18(33)28(5)23(29)22(35)24(40-14(2)32)27(4)16(15-6-7-37-11-15)8-21-30(27,28)41-21/h6-7,11,16-21,23-25,33-34,36H,8-10,12H2,1-5H3/t16-,17-,18+,19-,20+,21+,23-,24-,25+,26+,27+,28+,29+,30+/m0/

InChI key

NAHTXVIXCMUDLF-RFNFAWMESA-N

Biochem/physiol Actions

Botulinum neurotoxin inhibitor; Inhibitor of anthrax Lethal Toxin-induced cell death; exhibits potent antiviral activity against SARS-CoV-2, SFTSV, and bunyavirus.
Toosendanin is a natural occuring triterpenoid recently found to be an inhibitor of botulinum neurotoxin and also found to be an inhibitor of anthrax Lethal Toxin (LT)-induced cell death. Anthrax toxin is composed of three proteins: the toxin B subunit protective antigen (PA), a zinc metalloprotease lethal factor (LF), and an adenylate cyclase edema factor (EF). PA and EF combine to form edema toxin (ET) while PA and LF combine to form lethal toxin (LT), which induces rapid caspase-1 dependent cell death in macrophages. Toosendanin had an IC50 of 56 nM (32 ng/ml) against LT, likely by inhibiting endocytosis and endosomal acidification. Toosendanin has also been found to suppress proliferation and induce apoptosis in a variety of human cancer cells, possibly mediated through JNK signaling pathway.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service