Skip to Content
Merck
All Photos(1)

Key Documents

SML3462

Sigma-Aldrich

IAXO-102 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

IAXO 102 trifluoroacetate salt, Methyl 6-deoxy-6-amino-2,3-di-O-tetradecyl-α-D-glucopyranoside trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00
25 MG
$417.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$103.00
25 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C35H71NO5 · xC2HF3O2
Molecular Weight:
585.94 (free base basis)
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

SMILES string

O[C@H]1[C@@H]([C@H]([C@H](O[C@@H]1CN)OC)OCCCCCCCCCCCCCC)OCCCCCCCCCCCCCC.[xCF3CO2H]

Biochem/physiol Actions

IAXO-102 is a potent and selective toll-like receptor 4 (TLR4) antagonist that protect mice from LPS induced septic shock. IAXO-102 inhibits development of an experimental abdominal aortic aneurysm. It also prevents acute BBB disruption after experimental post-subarachnoid hemorrhage in mice.
Potent and selective toll-like receptor 4 (TLR4) antagonist that protect mice from LPS induced septic shock.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycolipids and benzylammonium lipids as novel antisepsis agents: synthesis and biological characterization.
Piazza M, Rossini C, Della Fiorentina S, Pozzi C, Comelli F, Bettoni I, Fusi P, Costa B, Peri F.
Journal of Medicinal Chemistry (2009)
A novel small molecule TLR4 antagonist (IAXO-102) negatively regulates non-hematopoietic toll like receptor 4 signalling and inhibits aortic aneurysms development.
Huggins C, Pearce S, Peri F, Neumann F, Cockerill G, Pirianov G.
Atherosclerosis (2015)
Selective Toll-Like Receptor 4 Antagonists Prevent Acute Blood-Brain Barrier Disruption After Subarachnoid Hemorrhage in Mice.
Okada T, Kawakita F, Nishikawa H, Nakano F, Liu L, Suzuki H.
Molecular Neurobiology (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service