Skip to Content
Merck
All Photos(1)

Key Documents

SML3460

Sigma-Aldrich

CB-103

≥97% (HPLC)

Synonym(s):

6-[4-(1,1-Dimethylethyl)phenoxy]-3-pyridinamine, CB 103, CB103

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$103.00
50 MG
$417.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$103.00
50 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C15H18N2O
CAS Number:
Molecular Weight:
242.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

, White to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Nc1cnc(cc1)Oc2ccc(cc2)C(C)(C)C

InChI

1S/C15H18N2O/c1-15(2,3)11-4-7-13(8-5-11)18-14-9-6-12(16)10-17-14/h4-10H,16H2,1-3H3

InChI key

WHIWGRCYMQLLAO-UHFFFAOYSA-N

Biochem/physiol Actions

CB-103 is an orally active Notch transcription activation inhibitor that targets RBPJ-Notch intracellular domain (NICD) transcription complex assembly. CB-103 exhibits antiproliferation efficacy against Notch-driven cancer growth in cultures (IC50 from 50 nM to 3 μM; patient-derived T-ALL cells positive for N1-ICD) and in tumor models in vivo (10-25 mg/kg qd or bid p.o. to mice with human T-ALL xenografts).
Orally active Notch transcription activation inhibitor with anti-cancer efficacy in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service