Skip to Content
Merck
All Photos(1)

Key Documents

SML3456

Sigma-Aldrich

LUF7244

≥98% (HPLC)

Synonym(s):

2-​[4-​(3-​Chlorobenzoyl)​phenoxy]​-​N-​3-​pyridinyl-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$575.00

$143.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$143.00
25 MG
$575.00

About This Item

Empirical Formula (Hill Notation):
C20H15ClN2O3
CAS Number:
Molecular Weight:
366.80
UNSPSC Code:
12352200
NACRES:
NA.77

$143.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NC1=CC=CN=C1)COC2=CC=C(C=C2)C(C3=CC=CC(Cl)=C3)=O

InChI key

KKKJYDOHVKIIQP-UHFFFAOYSA-N

Biochem/physiol Actions

LUF7244 is a potent and selective voltage-gated potassium 11.1 (Kv11.1) channels negative allosteric modulator/activator that inhibits early afterdepolarizations in vitro. It decreases the affinity of Kv11.1 for cisapride, astemizole, dofetilide and sertindole. LUF7244 averts dofetilide‐induced torsades de pointes arrhythmias in dogs with chronic atrioventricular block. LUF7244 in combination with dofetilide rescues both congenital and acquired Kv11.1 trafficking defects and produces functional Kv11.1 current in trafficking defective G601S-Kv11.1 cells.
Potent and selective voltage-gated potassium 11.1 (Kv11.1) channels negative allosteric modulator/activator that inhibits early afterdepolarizations

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muge Qile et al.
Molecular pharmacology, 97(6), 355-364 (2020-04-04)
Voltage-gated potassium 11.1 (Kv11.1) channels play a critical role in repolarization of cardiomyocytes during the cardiac action potential (AP). Drug-mediated Kv11.1 blockade results in AP prolongation, which poses an increased risk of sudden cardiac death. Many drugs, like pentamidine, interfere

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service