Skip to Content
Merck
All Photos(1)

Key Documents

SML3445

Sigma-Aldrich

VT104

≥98% (HPLC)

Synonym(s):

(S)-N-(l-(Pyridin-2-yl)ethyl)-5-(4-(trifluoromethyl)phenyl)-2-naphthamide, N-[(1S)-1-(2-Pyridinyl)ethyl]-5-[4-(trifluoromethyl)phenyl]-2-naphthalenecarboxamide, VT-104

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$194.00
25 MG
$783.00

$194.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$194.00
25 MG
$783.00

About This Item

Empirical Formula (Hill Notation):
C25H19F3N2O
CAS Number:
Molecular Weight:
420.43
UNSPSC Code:
12352200
NACRES:
NA.21

$194.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(C1=CC=C(C2=CC=CC3=CC(C(N[C@H](C4=NC=CC=C4)C)=O)=CC=C23)C=C1)F

InChI

1S/C25H19F3N2O/c1-16(23-7-2-3-14-29-23)30-24(31)19-10-13-22-18(15-19)5-4-6-21(22)17-8-11-20(12-9-17)25(26,27)28/h2-16H,1H3,(H,30,31)/t16-/m0/s1

InChI key

AAZUPSFRSHFTGV-INIZCTEOSA-N

Biochem/physiol Actions

Orally available pan-TEAD auto-palmitoylation inhibitor with anti-proliferation efficacy against mesothelioma with NF2 defect in cultures and in vivo.
VT104 is an orally available pan-TEAD auto-palmitoylation inhibitor (3 μM; using TEAD isotype-overexpressing HEK293T) that blocks TEAD-mediated gene transcription (IC50 = 10.4 nM) by preventing its association with YAP/TAZ. VT104 selectively inhibits the proliferation of mesothelioma cultures with NF2 mutation (NCI-H2051 IC50 = 31.6 nM), deletion (NCI-H2373 IC50 = 25.6 nM) or deficiency (NCI-H226 IC50 = 16.1 nM), but not those with wild-type NF2. Oral administration is efficacious against human mesothelioma NCI-H226 tumor growth in mice in vivo (1-3 mg/kg/day).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small Molecule Inhibitors of TEAD Auto-palmitoylation Selectively Inhibit Proliferation and Tumor Growth of NF2-deficient Mesothelioma
Tang TT, Konradi AW, Feng Y, Peng X, Ma M, Li J, Yu FX, Guan KL, Post L
Molecular Cancer Therapeutics, 20, 986-998 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service