Skip to Content
Merck
All Photos(1)

Key Documents

SML3392

Sigma-Aldrich

BC-11 hydrobromide

≥98% (HPLC)

Synonym(s):

Carbamimidothioic acid (4-boronophenyl)methyl ester hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$523.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$523.00

About This Item

Empirical Formula (Hill Notation):
C8H11BN2O2S·HBr
CAS Number:
Molecular Weight:
290.97
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

B(O)(O)c1ccc(cc1)CS\C(=N/[H])\N.Br

InChI

1S/C8H11BN2O2S.BrH/c10-8(11)14-5-6-1-3-7(4-2-6)9(12)13;/h1-4,12-13H,5H2,(H3,10,11);1H

InChI key

PAFZAMOVHIRQOD-UHFFFAOYSA-N

Biochem/physiol Actions

BC-11 is a potent and selective inhibitor of urokinase (urokinase-plasminogen activator, uPA). BC-11 inhibits clot lysis with no effect on clot formation. BC-11 exhibits anti-fibrotic activity and improves wound healing and scar quality after topical administration in mice.
potent and selective inhibitor of urokinase (urokinase-plasminogen activator, uPA)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Longo et al.
Molecules (Basel, Switzerland), 20(6), 9879-9889 (2015-06-02)
BC-11 is an easily synthesized simple thiouronium-substituted phenylboronic acid, which has been shown to be cytotoxic on triple negative MDA-MB231 breast cancer cells by inducing a perturbation of cell cycle when administered at a concentration equal to its ED50 at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service