Skip to Content
Merck
All Photos(1)

Key Documents

SML3345

Sigma-Aldrich

LSL 60101

≥98% (HPLC)

Synonym(s):

2-(1-Benzofuran-2-yl)-1H -imidazole, 2-(2-Benzofuranyl)-1H -imidazole (ACI), Garsevil, LSL60101

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$131.00
25 MG
$530.00

$131.00


Estimated to ship on17 April 2025



Select a Size

Change View
5 MG
$131.00
25 MG
$530.00

About This Item

Empirical Formula (Hill Notation):
C11H8N2O
CAS Number:
Molecular Weight:
184.19
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$131.00


Estimated to ship on17 April 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c(ncc1)c2[o]c3c(c2)cccc3

InChI

1S/C11H8N2O/c1-2-4-9-8(3-1)7-10(14-9)11-12-5-6-13-11/h1-7H,(H,12,13)

InChI key

VBFQIUOSXHUWCT-UHFFFAOYSA-N

Biochem/physiol Actions

LSL 60101 is an orally available, potent and selective I2 imidazoline receptor ligand that induces morphological/biochemical changes in astroglia and exhibits neuroprotective activity. LSL 60101 decreases neuroinflammation and reverses cognitive deficits and impaired social behavior in 5XFAD mice Alzheimer’s model.
Orally available, potent and selective I2 imidazoline receptor ligand that exhibits neuroprotective activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service