Skip to Content
Merck
All Photos(1)

Key Documents

SML3335

Sigma-Aldrich

DIPQUO

≥98% (HPLC)

Synonym(s):

6,​8-​Dimethyl-​3-​(4-​phenyl-​1H-​imidazol-​5-​yl)​-2(1H)​-​quinolinone, 6,8-Dimethyl-3-(4-phenyl-1H-imidazol-5-yl)quinolin-2(1H)-one, RU 0226606

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$523.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$523.00

About This Item

Empirical Formula (Hill Notation):
C20H17N3O
CAS Number:
Molecular Weight:
315.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NC2=C(C)C=C(C)C=C2C=C1C3=C(C4=CC=CC=C4)N=CN3

Biochem/physiol Actions

Activator of the bone marker alkaline phosphatase (ALP) that promotes osteogenesis; promotes activation of the β isoform of p38 MAP kinase
DIPQUO is a potent activator of the bone marker alkaline phosphatase (ALP) that promotes osteogenesis in human MSCs and zebrafish. DIPQUO potently induces osteoblast differentiation. It promotes bone formation and regeneration in zebra fish. It appears that DIPQUO promotes activation of the β isoform of p38 MAP kinase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a Small Molecule Promoting Mouse and Human Osteoblast Differentiation via Activation of p38 MAPK-?
Cell Chemical Biology, 26 (2019)
Brandoch Cook et al.
The Journal of biological chemistry, 296, 100696-100696 (2021-04-26)
Bone fractures are common impact injuries typically resolved through natural processes of osteogenic regeneration and bone remodeling, restoring the biological and mechanical function. However, dysfunctionality in bone healing and repair often arises in the context of aging-related chronic disorders, such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service