Skip to Content
Merck
All Photos(1)

Documents

SML3286

Sigma-Aldrich

cUMP sodium salt

≥98% (HPLC)

Synonym(s):

3’,5’-cyclic uridine monophosphate sodium salt, CID 3081385 sodium, Uridine cyclic 3′,5′-monophosphate monosodium salt, cyclic UMP sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O8P·Na
CAS Number:
Molecular Weight:
328.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@H]1[C@@](N2C(NC(C=C2)=O)=O)([H])O[C@@]3([H])[C@@]1([H])OP(OC3)(O[Na])=O

InChI

1S/C9H11N2O8P.Na/c12-5-1-2-11(9(14)10-5)8-6(13)7-4(18-8)3-17-20(15,16)19-7;/h1-2,4,6-8,13H,3H2,(H,15,16)(H,10,12,14);/q;+1/p-1

InChI key

APYWIKXPIFYSBG-UHFFFAOYSA-M

General description

3’,5’-cyclic uridine monophosphate (cUMP) is a cyclic pyrimidine nucleotide. cUMP (3’,5’-cyclic uridine monophosphate) sodium salt is a secondary messenger functioning in bacterial immunity against viruses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sander de Kivit et al.
Nature metabolism, 2(10), 1046-1061 (2020-09-23)
Following activation, conventional T (Tconv) cells undergo an mTOR-driven glycolytic switch. Regulatory T (Treg) cells reportedly repress the mTOR pathway and avoid glycolysis. However, here we demonstrate that human thymus-derived Treg (tTreg) cells can become glycolytic in response to tumour
Nicole Blaquiere et al.
Journal of medicinal chemistry, 61(15), 6801-6813 (2018-06-26)
NF-κB-inducing kinase (NIK) is a protein kinase central to the noncanonical NF-κB pathway downstream from multiple TNF receptor family members, including BAFF, which has been associated with B cell survival and maturation, dendritic cell activation, secondary lymphoid organ development, and
Weisong Duan et al.
Molecular neurobiology, 58(5), 2435-2446 (2021-01-14)
TANK-binding kinase1 (TBK1) haploinsufficiency has been shown to cause both amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD); however, the mechanism is unclear. Myeloid Tbk1 knockout mice (Tbk1-LKO mice) were established and motor function and pathological analyses were also performed.
Jieqiong Zhang et al.
Cell reports, 27(9), 2679-2689 (2019-05-30)
Cellular inhibitor of apoptosis proteins cIAP1 and cIAP2 ubiquitinate nuclear factor κB (NF-κB)-inducing kinase (NIK) to suppress non-canonical NF-κB signaling and substrates such as receptor interacting protein kinase 1 (RIPK1) to promote cell survival. We investigate how these functions contribute to
Jiang Wang et al.
Pathogens and disease, 78(7) (2020-08-30)
Periodontitis is an inflammatory disease that causes damages to periodontium and alveolar bone. Overactivation and formation of osteoclasts can cause bone destruction, which contributes to periodontitis development. Receptor activator of nuclear factor κB ligand (RANKL)-mediated NF-κB signaling plays an essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service