Skip to Content
Merck
All Photos(1)

Key Documents

SML3239

Sigma-Aldrich

BI-5026

≥98% (HPLC)

Synonym(s):

3-Amino-N,N-dimethyl-6-(4-(methylsulfonyl)piperidin-1-yl)-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide, BI 5026, BI 605906 inactive control, BI 605906 negative control, BI-605906 inactive control, BI5026, BI605906 inactive control, BI605906 negative control

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$120.00
25 MG
$484.00

$120.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$120.00
25 MG
$484.00

About This Item

Empirical Formula (Hill Notation):
C17H21F3N4O3S2
Molecular Weight:
450.50
UNSPSC Code:
12352107
NACRES:
NA.77

$120.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(SC2=C1C(C(F)(F)F)=CC(N3CCC(CC3)S(C)(=O)=O)=N2)C(N(C)C)=O

InChI

1S/C17H21F3N4O3S2/c1-23(2)16(25)14-13(21)12-10(17(18,19)20)8-11(22-15(12)28-14)24-6-4-9(5-7-24)29(3,26)27/h8-9H,4-7,21H2,1-3H3

InChI key

HHYGVXHXNNIDJF-UHFFFAOYSA-N

Biochem/physiol Actions

BI-5026 is an inactive analog of and negative control for the sective, ATP-competitive IκB kinase β (IKK2, IKKβ, IKK-beta) inhibitor BI605906
Inactive analog of and negative control for the sective, ATP-competitive IκB kinase β (IKK2, IKKβ, IKK-beta) inhibitor BI605906.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service