Skip to Content
Merck
All Photos(1)

Key Documents

SML3185

Sigma-Aldrich

CGP 48664 dihydrochloride

≥98% (HPLC)

Synonym(s):

(E)-2-(4-carbamimidoyl-2,3-dihydro-1H-inden-1-ylidene)hydrazine-1-carboximidamide dihydrochloride, (E)-2-[4-(Aminoiminomethyl)-2,3-dihydro-1H-inden-1-ylidene]hydrazinecarboximidamide dihydrochloride, 4-Amidino-1-indanone-2′-amidinohydrazone dihydrochloride, 4-Amidinoindan-1-one-2?-amidinohydrazone dihydrochloride, CGP 48664A dihydrochloride, CGP48664 dihydrochloride, CMA 48664 dihydrochloride, SAM486A dihydrochloride, Sardomozide dihydrochlorid, Sardomozide dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$226.00
25 MG
$906.00

$226.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$226.00
25 MG
$906.00

About This Item

Empirical Formula (Hill Notation):
C11H14N6 · 2HCl
CAS Number:
Molecular Weight:
303.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$226.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

NC(C1=CC=CC2=C1CC/C2=N\NC(N)=N)=N.Cl.Cl

InChI

1S/C11H14N6/c12-10(13)8-3-1-2-7-6(8)4-5-9(7)16-17-11(14)15/h1-3H,4-5H2,(H3,12,13)(H4,14,15,17)/b16-9+

InChI key

CYPGNVSXMAUSJY-CXUHLZMHSA-N

Biochem/physiol Actions

CGP 48664 (SAM486A; Sardomozide) is a polyamine (PA) biosynthesis inhibitor (73% and 97% downregulation of murine leukemia L1210 cellular spermidine and spermine, respectively, by 0.5 μM Sardomozide in 4 days) that targets the rate-limiting enzyme S-adenosylmethionine decarboxylase (AdoMetDC, SAMDC, AMD1) in a potent (IC50 = 5 nM against rat AMD1) and selective manner with little or no inhibitory efficacy toward diamine oxidase or ornithine decarboxylase (IC50 = 18 μM and >50 μM against rat DAO and ODC, respectively). CGP 48664 effectively inhibits the proliferation of murine (GI50 = 0.5 μM against L1210 in 4 days) and human cancer cell lines in cultures (GI50 from 0.26 to 0.6 μM in 5 days; GI50 = 0.8 μM against human melanoma SK MEL-24 in 10 days) and suppresses the growth of murine and human tumor xenografts in mice in vivo (ED50 ≤0.5 mg/kg i.p.). CGP 48664 (Sardomozide) potently inhibits growth of PFA ependymoma cells.
Potent and selective S-adenosylmethionine decarboxylase (AdoMetDC, SAMDC, AMD1) inhibitor with anti-cancer efficacy both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service