Skip to Content
Merck
All Photos(1)

Key Documents

SML3176

Sigma-Aldrich

WEHI-539 hydrochloride

≥98% (HPLC)

Synonym(s):

(E)-5-(3-(4-(Aminomethyl)phenoxy)propyl)-2-(8-(2-(benzo[d]thiazol-2-yl)hydrazono)-5,6,7,8-tetrahydronaphthalen-2-yl)thiazole-4-carboxylic acid hydrochloride, 5-[3-[4-(Aminomethyl)phenoxy]propyl]-2-[8-[2-(2-benzothiazolyl)hydrazinylidene]-5,6,7,8-tetrahydro-2-naphthalenyl]-4-thiazolecarboxylic acid hydrochloride, WEHI 539 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$518.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$518.00

About This Item

Empirical Formula (Hill Notation):
C31H29N5O3S2 · xHCl
Molecular Weight:
583.72 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c(nc(c1CCCOc6ccc(cc6)CN)C(=O)O)c2cc3c(cc2)CCC=C3NNc4[s]c5c(n4)cccc5

InChI

1S/C31H29N5O3S2/c32-18-19-10-14-22(15-11-19)39-16-4-9-27-28(30(37)38)34-29(40-27)21-13-12-20-5-3-7-24(23(20)17-21)35-36-31-33-25-6-1-2-8-26(25)41-31/h1-2,6-8,10-15,17,35H,3-5,9,16,18,32H2,(H,33,36)(H,37,38)

InChI key

BDQRJRWAPLGIFF-UHFFFAOYSA-N

Biochem/physiol Actions

An apoptosis inducer that selectively targets the anti-apoptotic Bcl-2 family member Bcl-XL with high affinity.
WEHI-539 is an apoptosis inducer that selectively targets the anti-apoptotic Bcl-2 family member Bcl-XL with high affinity (KD = 0.6 nM; 500-fold over Bcl-2 and 400-fold over Bcl-W, Mcl-1 and A1) in a manner distinct from that of ABT-737. WEHI blocks Bcl-XL from targeting the BH3 domain of the pro-apoptotic family member Bim with high potency (IC50 = 1.1 nM), exhibiting no inhibitory potency against Mcl-1-BimBH3 interaction (IC50 >10 μM). WEHI-539 is shown to induce apoptosis in human and murine cultures by selectively antagonizing Bcl-XL prosurvival activity (EC50 <10 nM/Mcl-1-knockout MEF, <0.5 nM/mouse platelets, and <32 nM/human platelets).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service