Skip to Content
Merck
All Photos(1)

Key Documents

SML3132

Sigma-Aldrich

Tivozanib

≥98% (HPLC)

Synonym(s):

1-[2-Chloro-4-(6,7-dimethoxyquinolin-4-yl)oxyphenyl]-3-(5-methyl-1,2-oxazol-3-yl)urea, AV-951, KRN951, N-[2-Chloro-4-[(6,7-dimethoxy-4-quinolinyl)oxy]phenyl]-N?-(5-methyl-3-isoxazolyl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$180.00
50 MG
$578.00

$180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$180.00
50 MG
$578.00

About This Item

Empirical Formula (Hill Notation):
C22H19ClN4O5
CAS Number:
Molecular Weight:
454.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$180.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=C(Cl)C=C(C=C1)OC2=C3C=C(C(OC)=CC3=NC=C2)OC)NC4=NOC(C)=C4

InChI

1S/C22H19ClN4O5/c1-12-8-21(27-32-12)26-22(28)25-16-5-4-13(9-15(16)23)31-18-6-7-24-17-11-20(30-3)19(29-2)10-14(17)18/h4-11H,1-3H3,(H2,25,26,27,28)

InChI key

SPMVMDHWKHCIDT-UHFFFAOYSA-N

Biochem/physiol Actions

Tivozanib (KRN951) is an oral, next-generation vascular endothelial growth factor receptor (VEGFR) tyrosine kinase inhibitor (TKI). Tivozanib is a potent, selective inhibitor of VEGFRs 1, 2, and 3 with a long half-life designed to improve efficacy and tolerability.
orally active, potent, selective inhibitor of VEGFRs 1, 2, and 3 with a long half-life designed to improve efficacy and tolerability

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service