Skip to Content
Merck
All Photos(1)

Key Documents

SML3116

Sigma-Aldrich

BMS-378806

≥98% (HPLC)

Synonym(s):

1-[(2R )-4-Benzoyl-2-methyl-1-piperazinyl]-2-(4-methoxy-1H -pyrrolo[2,3-b ]pyridin-3-yl)-1,2-ethanedione, 4-Benzoyl-1-[(4-methoxy-1H- pyrrolo[2,3-b]pyridin-3-yl)oxoacetyl]-2-(R)-methylpiperazine, BMS 378806, BMS 806, BMS-806, BMS378806, BMS806

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$156.00
50 MG
$625.00

$156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$156.00
50 MG
$625.00

About This Item

Empirical Formula (Hill Notation):
C22H22N4O4
CAS Number:
Molecular Weight:
406.43
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

$156.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1[C@@H](CN(CC1)C(C2=CC=CC=C2)=O)C)C(C3=CNC4=NC=CC(OC)=C34)=O

InChI

1S/C22H22N4O4/c1-14-13-25(21(28)15-6-4-3-5-7-15)10-11-26(14)22(29)19(27)16-12-24-20-18(16)17(30-2)8-9-23-20/h3-9,12,14H,10-11,13H2,1-2H3,(H,23,24)/t14-/m1/s1

InChI key

OKGPFTLYBPQBIX-CQSZACIVSA-N

General description

BMS-378806 is an indole-based compound that exhibits improved antiviral potency, and pharmaceutical and pharmacokinetic activities.[1] BMS-378806 is inactive against HIV-2 and Simian immunodeficiency virus (SIV) and a few other viruses.[2]

Biochem/physiol Actions

BMS-378806 (BMS-806) is a potent and selective HIV-1 attachment inhibitor that blocks HIV glycoprotein 120 (gp120) from interacting with host T-cell surface glycoprotein CD4 (IC50 = 100 nM against CD4 for binding gp120; CYP1A2/2C9/2C19/2D6/3A4 IC50 >23 μM) without affecting HIV integrase, protease, or reverse transcriptase activity. BMS-806 effectively prevents various HIV-1 isolates from replication (EC50 from 0.85 to 26.5 nM in MT-2 and PM1 cultures; host cytotoxicity IC50 >300 μM) without affecting HIV-2, SIV, MuLV, RSV, HCMV, BVDV, VSV, or influenza virus.
Potent and selective HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions and protects against HIV-1 infection in host cultures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service