Skip to Content
Merck
All Photos(1)

Key Documents

SML3107

Sigma-Aldrich

Glycycoumarin

≥98% (HPLC)

Synonym(s):

3-(2,4-Dihydroxy-phenyl)-7-hydroxy-5-methoxy-6-(3-methyl-but-2-enyl)-1-benzopyran-2-one, 3-(2,4-Dihydroxyphenyl)-7-hydroxy-5-methoxy-6-(3-methyl-2-buten-1-yl)-2H-1-benzopyran-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$144.00
25 MG
$583.00

$144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$144.00
25 MG
$583.00

About This Item

Empirical Formula (Hill Notation):
C21H20O6
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$144.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[o]1c2c(c(c(c(c2)O)CC=C(C)C)OC)cc([c]1=O)c3c(cc(cc3)O)O

InChI

1S/C21H20O6/c1-11(2)4-6-14-18(24)10-19-16(20(14)26-3)9-15(21(25)27-19)13-7-5-12(22)8-17(13)23/h4-5,7-10,22-24H,6H2,1-3H3

InChI key

NZYSZZDSYIBYLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glycycoumarin is an isoflavone from vegetable roots including licorice that exhibits potent hepatoprotective effects. Glycycoumarin protects hepatocytes through activation of autophagy and mitigation of ER stress/GSK-3 –JNK/CHOP-mitochondria cascade activation. Also, it appears to alleviate acetaminophen-induced liver injury through sustained autophagy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service