Skip to Content
Merck
All Photos(1)

Key Documents

SML3057

Sigma-Aldrich

Indacaterol

≥98% (HPLC)

Synonym(s):

5-[(1R)-2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-2(1H)-quinolinone, QAB-149, R)-5-[2-(5,6-diethylindan-2-ylamino)-1-hydroxyethyl]-8-hydroxy-1H-quinolin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$105.00
50 MG
$328.00

$105.00


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$105.00
50 MG
$328.00

About This Item

Empirical Formula (Hill Notation):
C24H28N2O3
CAS Number:
Molecular Weight:
392.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$105.00


Estimated to ship on06 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@H](CNC1CC(C=C(CC)C(CC)=C2)=C2C1)C3=CC=C(O)C(N4)=C3C=CC4=O

InChI

1S/C24H28N2O3/c1-3-14-9-16-11-18(12-17(16)10-15(14)4-2)25-13-22(28)19-5-7-21(27)24-20(19)6-8-23(29)26-24/h5-10,18,22,25,27-28H,3-4,11-13H2,1-2H3,(H,26,29)/t22-/m0/s1

InChI key

QZZUEBNBZAPZLX-QFIPXVFZSA-N

Biochem/physiol Actions

Indacaterol is an ultra-long-acting beta-adrenoceptor agonist. It is used as bronchodilator.
Ultra-long-acting beta-adrenoceptor agonist; bronchodilator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radhika Joshi et al.
Molecular pharmacology, 96(1), 56-72 (2019-05-01)
The effects of phosphodiesterase (PDE) 4 inhibitors on gene expression changes in BEAS-2B human airway epithelial cells are reported and discussed in relation to the mechanism(s) of action of roflumilast in chronic obstructive pulmonary disease (COPD). Microarray-based gene expression profiling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service