Skip to Content
Merck
All Photos(1)

Key Documents

SML3046

Sigma-Aldrich

Saxagliptin hydrate

≥98% (HPLC)

Synonym(s):

(S)-3-Hydroxyadamantylglycine-L-cis-4,5-methanoprolinenitrile hydrate, BMS 477118, BMS 477118-11, BMS-477118, BMS-477118-11, BMS477118, BMS477118-11

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$137.00
50 MG
$553.00

$137.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$137.00
50 MG
$553.00

About This Item

Empirical Formula (Hill Notation):
C18H25N3O2 · H2O
CAS Number:
Molecular Weight:
333.43
UNSPSC Code:
12352200
NACRES:
NA.77

$137.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OC12CC([C@@H](C(N3[C@@H](C[C@]4([C@@]3(C4)[H])[H])C#N)=O)N)(CC(C5)C2)CC5C1.O

InChI

1S/C18H25N3O2.H2O/c19-8-13-2-12-3-14(12)21(13)16(22)15(20)17-4-10-1-11(5-17)7-18(23,6-10)9-17;/h10-15,23H,1-7,9,20H2;1H2/t10?,11?,12-,13+,14+,15-,17?,18?;/m1./s1

InChI key

AFNTWHMDBNQQPX-NHKADLRUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Orally active, highly potent & long-acting dipeptidyl peptidase IV (DPP-IV) inhibitor with therapeutic efficacy against type II diabetes in vivo.
Saxagliptin (BMS-477118) is an orally active, highly potent and long-acting dipeptidyl peptidase IV inhibitor (human DPP-IV Ki = 0.9 nM; plasma DDP-IV inhibition = 87% 0.5-4h post 4 μmol/kg po. in normal rats) with therapeutic efficacy against type 2 diabetes in vivo (plasma glucose clearance = 28/43/61% post 0.3/1.0/3.0 μmol/kg po. in Zucker fa/fa rats).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suat Tekin et al.
Archives of physiology and biochemistry, 1-11 (2020-01-26)
Saxagliptin is an effective and selective dipeptidyl peptidase-4 (DPP-4) inhibitor. This study was designed to determine possible protective effects of saxagliptin against damage caused by renal ischaemia/reperfusion (I/R) in rats. In this study, 40 rats were divided into 4 groups

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service