Skip to Content
Merck
All Photos(1)

Key Documents

SML3026

Sigma-Aldrich

BAY-786

≥98% (HPLC)

Synonym(s):

(2R)-N-[[3-Chloro-5-(trifluoromethyl)-2-pyridyl]methyl]-2-cyclopropyl-1-(4-fluorophenyl)sulfonyl-1′,1′-dioxo-spiro[indoline-3,4′-thiane]-5-carboxamide, (2R)-N-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]methyl]-2-cyclopropyl-1-[(4-fluorophenyl)sulfonyl]-1,2,2,3,5,6-hexahydro-spiro[3H-indole-3,4-[4H]thiopyran]-5-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$207.00
25 MG
$829.00

$207.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$207.00
25 MG
$829.00

About This Item

Empirical Formula (Hill Notation):
C29H26ClF4N3O5S2
CAS Number:
Molecular Weight:
672.11
UNSPSC Code:
12352200
NACRES:
NA.77

$207.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 105 to 125°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC2=C(N(S(=O)(C3=CC=C(F)C=C3)=O)[C@H](C4CC4)C2(CC5)CCS5(=O)=O)C=C1)NCC6=NC=C(C(F)(F)F)C=C6Cl

InChI key

PZGSYNNVPNLHQG-AREMUKBSSA-N

Biochem/physiol Actions

BAY-786 is a negative control for an orally available, non-toxic, potent, and selective GnRH-R antagonist BAY-784.
negative control for an orally available, non-toxic, potent, and selective GnRH-R antagonist BAY-784

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olaf Panknin et al.
Journal of medicinal chemistry, 63(20), 11854-11881 (2020-09-23)
The growth of uterine fibroids is sex hormone-dependent and commonly associated with highly incapacitating symptoms. Most treatment options consist of the control of these hormonal effects, ultimately blocking proliferative estrogen signaling (i.e., oral contraceptives/antagonization of human gonadotropin-releasing hormone receptor [hGnRH-R]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service