Skip to Content
Merck
All Photos(1)

Key Documents

SML3021

Sigma-Aldrich

ADLumin-1

≥98% (HPLC)

Synonym(s):

6-((1E,3E)-4-(4-(dimethylamino)phenyl)buta-1,3-dien-1-yl)-2-methylimidazo[1,2-a]pyrazin-3(7H)-one, 6-[4-[4-(Dimethylamino)phenyl]-1,3-butadien-1-yl]-2-methyl-imidazo[1,2-a]pyrazin-3(7H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$518.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$518.00

About This Item

Empirical Formula (Hill Notation):
C19H20N4O
CAS Number:
Molecular Weight:
320.39
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

H2O: 0.1 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

CN(C1=CC=C(C=C1)/C=C/C=C/C2=CN3C(N=C(C3=O)C)=CN2)C

Biochem/physiol Actions

ADLumin-1 brain blood barrier penetrant smart chemiluminescence probe for Aβ (amyloid beta) species. Upon binding to Aβ, ADLumin-1 chemiluminescence dramatically increases and its peak shifts from 590 nm to 540 nm. ADLumin-1 is suitable for in vivo dually-amplify signal via chemiluminescence resonance energy transfer (DAS-CRET) with CRANAD-3 a smart NIRF (near infrared fluorescence imaging) probe for Aβs. Apparently both probes bind to Aβ aggregates in close proximity.
brain blood barrier penetrant smart chemiluminescence probe for Aβ (amyloid beta) species

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Yang et al.
Nature communications, 11(1), 4052-4052 (2020-08-15)
Turn-on fluorescence imaging is routinely studied; however, turn-on chemiluminescence has been rarely explored for in vivo imaging. Herein, we report the design and validation of chemiluminescence probe ADLumin-1 as a turn-on probe for amyloid beta (Aβ) species. Two-photon imaging indicates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service