Skip to Content
Merck
All Photos(1)

Key Documents

SML3020

Sigma-Aldrich

Taltirelin acetate

≥98% (HPLC)

Synonym(s):

(4S)-Hexahydro-1-methyl-2,6-dioxo-4-pyrimidinecarbonyl-L-histidyl-L-prolinamide, (S)-N-((S)-1-((S)-2-Carbamoylpyrrolidin-1-yl)-3-(1H-imidazol-4-yl)-1-oxopropan-2-yl)-1-methyl-2,6-dioxohexahydropyrimidine-4-carboxamide, (S)-N-[(Hexahydro-1-methyl-2,6-dioxo-4-pyrimidinyl)carbonyl]-L-histidyl-L-prolinamide, TA 0910, TA-0910, TA0910

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$120.00
50 MG
$484.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$120.00
50 MG
$484.00

About This Item

Empirical Formula (Hill Notation):
C17H23N7O5 · xC2H4O2
Molecular Weight:
405.41 (free base basis)
UNSPSC Code:
12352107
NACRES:
NA.77

$120.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

Taltirelin (TA-0910) is a thyrotropin-releasing hormone (TRH) mimetic with greatly enhanced brain-penetration, oral activity and stability. Taltirelin shortens the duration of pentobarbital-induced sleep (0.3-3 mg/kg po. or 0.03-0.3 mg/kg iv.), antagonizes reserpine-induced hypothermia (1 or 3 mg/kg po. or 0.1-0.3 mg/kg ip.), and enhances the spontaneous motor activity (30 mg/kg po.) in mice in vivo, while TRH produces these effects at about 100 times higher doses.
Thyrotropin-releasing hormone (TRH) mimetic with greatly enhanced brain-penetration, oral activity, stability and in vivo efficacy than TRH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yamamura et al.
Japanese journal of pharmacology, 53(4), 451-461 (1990-08-01)
Effects of orally administered TA-0910, a new thyrotropin-releasing hormone (TRH) analog, on the central nervous system (CNS) were investigated and compared with those of TRH. TA-0910 shortened the duration of pentobarbital-induced sleep and antagonized reserpine-induced hypothermia at 0.3 mg/kg or
John P Dougherty et al.
Behavioural brain research, 360, 255-261 (2018-12-12)
Fatigue is a common symptom in many diseases and disorders and can reduce quality of life, yet lacks an adequate pharmacological intervention. To identify and develop such interventions, and to better understand fatigue, additional preclinical research is necessary. However, despite
Cong Zheng et al.
Frontiers in cellular neuroscience, 12, 485-485 (2019-01-09)
Dopaminergic neurons loss is one of the main pathological characters of Parkinson's disease (PD), while no suitable neuroprotective agents have been in clinical use. Thyrotropin-releasing hormone (TRH) and its analogs protect neurons from ischemia and various cytotoxins, but whether the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service