Skip to Content
Merck
All Photos(1)

Key Documents

SML3007

Sigma-Aldrich

H3B-120

≥98% (HPLC)

Synonym(s):

N1-Methyl-N4-(4-methyl-2-thiazolyl)-N1-(phenylmethyl)-1,4-piperidinedicarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$171.00
25 MG
$553.00

$171.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$171.00
25 MG
$553.00

About This Item

Empirical Formula (Hill Notation):
C19H24N4O2S
CAS Number:
Molecular Weight:
372.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$171.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1CCN(CC1)C(N(C)CC2=CC=CC=C2)=O)NC3=NC(C)=CS3

InChI

1S/C19H24N4O2S/c1-14-13-26-18(20-14)21-17(24)16-8-10-23(11-9-16)19(25)22(2)12-15-6-4-3-5-7-15/h3-7,13,16H,8-12H2,1-2H3,(H,20,21,24)

InChI key

ZEVOZXRSCSSPAO-UHFFFAOYSA-N

Biochem/physiol Actions

H3B-120 is a cell penetrant, potent and selective allosteric inhibitor of the carbamoyl phosphate synthetase 1 (CPS1) that blocks ATP hydrolysis in the first step of carbamoyl phosphate synthesis
cell penetrant, potent and selective allosteric inhibitor of the carbamoyl phosphate synthetase 1 (CPS1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shihua Yao et al.
Cell chemical biology, 27(3), 259-268 (2020-02-06)
Carbamoyl phosphate synthetase 1 (CPS1) catalyzes the first step in the ammonia-detoxifying urea cycle, converting ammonia to carbamoyl phosphate under physiologic conditions. In cancer, CPS1 overexpression supports pyrimidine synthesis to promote tumor growth in some cancer types, while in others

Questions

  1. Hello, I would like to know how to conserve the product H3B-120 - SML3007-5MG , after suspension in DMSO at the concentration 2mg/ml. Is it possible to do aliquots and keep it at -20°C? Thank you for your help

    1 answer
    1. Unfortunately, long-term solution stability studies have not been performed on this product. Other sources suggest that stock solutions in DMSO may be stable for up to 1 year at -20°C in frozen aliquots. This information has not been validated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service