Skip to Content
Merck
All Photos(1)

Key Documents

SML2990

Sigma-Aldrich

dCeMM2

≥98% (HPLC)

Synonym(s):

2-((5H-[1,2,4]Triazino[5,6-b]indol-3-yl)thio)-N-(5-chloropyridin-2-yl)acetamide, N-(5-Chloro-2-pyridinyl)-2-(5H-1,2,4-triazino[5,6-b]indol-3-ylthio)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$137.00
25 MG
$691.00

$137.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$137.00
25 MG
$691.00

About This Item

Empirical Formula (Hill Notation):
C16H11ClN6OS
CAS Number:
Molecular Weight:
370.82
UNSPSC Code:
12352200
NACRES:
NA.77

$137.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=NC=C(Cl)C=C1)CSC2=NC3=C(C4=C(N3)C=CC=C4)N=N2

InChI

1S/C16H11ClN6OS/c17-9-5-6-12(18-7-9)20-13(24)8-25-16-21-15-14(22-23-16)10-3-1-2-4-11(10)19-15/h1-7H,8H2,(H,18,20,24)(H,19,21,23)

InChI key

VFWVTIHYVUKNGR-UHFFFAOYSA-N

Biochem/physiol Actions

cell penetrant molecular glue degrader that induce ubiquitination and degradation of cyclin K by inciting an interaction of CDK12–cyclin K with a CRL4B ligase
dCeMM2 is a cell penetrant molecular glue degrader that induce ubiquitination and degradation of cyclin K by inciting an interaction of CDK12–cyclin K with a CRL4B ligase complex. dCeMM2 induced proximity between CDK12–cyclin K and the CRL complex adaptor protein DDB1 that causes ubiquitination and proteasomal degradation of cyclin K.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Mayor-Ruiz et al.
Nature chemical biology, 16(11), 1199-1207 (2020-08-05)
Targeted protein degradation is a new therapeutic modality based on drugs that destabilize proteins by inducing their proximity to E3 ubiquitin ligases. Of particular interest are molecular glues that can degrade otherwise unligandable proteins by orchestrating direct interactions between target

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service