Skip to Content
Merck
All Photos(1)

Key Documents

SML2973

Sigma-Aldrich

MCA-AVLQSGFR-Lys(Dnp)-Lys-NH2 trifluoroacetate

≥95% (HPLC)

Synonym(s):

N-[2-(7-methoxy-2-oxo-2H-1-benzopyran-4-yl)acetyl]-L-alanyl-L-valyl-L-leucyl-L-glutaminyl-L-serylglycyl-L-phenylalanyl-L-arginyl-N6-(2,4-dinitrophenyl)-L-lysyl-L-Lysinamide trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$169.00
5 MG
$683.00

$169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$169.00
5 MG
$683.00

About This Item

Empirical Formula (Hill Notation):
C69H99N19O20 · xC2HF3O2
CAS Number:
Molecular Weight:
1514.64 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$169.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

film or powder

color

white to yellow

storage temp.

−20°C

Biochem/physiol Actions

FRET substrate suitable to measure protease activity of COVID-19 virus Mpro, SARS-CoV Mpro, and some other viruses
MCA-AVLQSGFR-Lys(Dnp)-Lys-NH2 is a fluorescence resonance energy transfer (FRET) substrate derived from the N-terminal auto-cleavage fragment of the SARS-CoV Mpro viral protease ((excitation wavelengths 320 nm and emission wavelengths 405 nm). MCA-AVLQSGFR-Lys(Dnp)-Lys-NH2 is suitable to measure protease activity of COVID-19 virus Mpro, SARS-CoV Mpro, and other viruses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyu Xue et al.
Journal of molecular biology, 366(3), 965-975 (2006-12-27)
The viral proteases have proven to be the most selective and useful for removing the fusion tags in fusion protein expression systems. As a key enzyme in the viral life-cycle, the main protease (M(pro)) is most attractive for drug design
Fenghua Wang et al.
Scientific reports, 6, 22677-22677 (2016-03-08)
First identified in The Netherlands in 2004, human coronavirus NL63 (HCoV-NL63) was found to cause worldwide infections. Patients infected by HCoV-NL63 are typically young children with upper and lower respiratory tract infection, presenting with symptoms including croup, bronchiolitis, and pneumonia.
Zhenming Jin et al.
Nature, 582(7811), 289-293 (2020-04-10)
A new coronavirus, known as severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), is the aetiological agent responsible for the 2019-2020 viral pneumonia outbreak of coronavirus disease 2019 (COVID-19)1-4. Currently, there are no targeted therapeutic agents for the treatment of this disease, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service