Skip to Content
Merck
All Photos(1)

Key Documents

SML2968

Sigma-Aldrich

JA2131

≥98% (HPLC)

Synonym(s):

1,3,6,9-Tetrahydro-1,3-dimethyl-8-[[2-(4-morpholinyl)ethyl]thio]-6-thioxo-2H-purin-2-one, NSC 98003, NSC98003

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$238.00
25 MG
$761.00

$238.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$238.00
25 MG
$761.00

About This Item

Empirical Formula (Hill Notation):
C13H19N5O2S2
CAS Number:
Molecular Weight:
341.45
UNSPSC Code:
12352200
NACRES:
NA.77

$238.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S(CCN3CCOCC3)c1[nH]c2c(n1)N(C(=O)N(C2=S)C)C

InChI

1S/C13H19N5O2S2/c1-16-10-9(11(21)17(2)13(16)19)14-12(15-10)22-8-5-18-3-6-20-7-4-18/h3-8H2,1-2H3,(H,14,15)

InChI key

XHNSOGDMSSHQFY-UHFFFAOYSA-N

Biochem/physiol Actions

JA2131 is a cell penetrant, potent and selective poly(ADP-ribose) glycohydrolase (PARG) inhibitor that selectively kills cancer cells. JA2131 induces PARP1 hyper-PARylation and sensitizes cancer cells to radiation-induced DNA damage. JA2131 induces replication fork defects.
cell penetrant, potent and selective poly(ADP-ribose) glycohydrolase (PARG) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry H Houl et al.
Nature communications, 10(1), 5654-5654 (2019-12-13)
Poly(ADP-ribose)ylation (PARylation) by PAR polymerase 1 (PARP1) and PARylation removal by poly(ADP-ribose) glycohydrolase (PARG) critically regulate DNA damage responses; yet, conflicting reports obscure PARG biology and its impact on cancer cell resistance to PARP1 inhibitors. Here, we found that PARG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service