Skip to Content
Merck
All Photos(1)

Key Documents

SML2912

Sigma-Aldrich

UNC4976 Trifluoroacetate

≥95% (HPLC)

Synonym(s):

Methyl N6-(bicyclo[2.2.1]heptan-2-yl)-N2-(4-(tert-butyl)benzoyl)-L-phenylalanyl-L-alanyl-L-leucyl-N6-methyl-L-lysyl-L-serinate trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$169.00
25 MG
$593.00

$169.00


Check Cart for Availability


Select a Size

Change View
5 MG
$169.00
25 MG
$593.00

About This Item

Empirical Formula (Hill Notation):
C47H70N6O8 · xC2HF3O2
Molecular Weight:
847.09 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$169.00


Check Cart for Availability

Quality Level

Assay

≥95% (HPLC)

form

film

color

white to beige

shipped in

wet ice

storage temp.

−20°C

SMILES string

N([C@@H](CO)C(=O)OC)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)c4ccc(cc4)C(C)(C)C)Cc3ccccc3)C)CC(C)C)CCCCN(C1C2CC(C1)CC2)C

InChI key

UKWNWTBRKZUHGX-GKUWNYOMSA-N

Biochem/physiol Actions

UNC4976 is a cell penetrant and potent positive allosteric modulator (PAM) of CBX7 chromodomain of polycomb repressive complex 1 that enhances CBX7 chromodomain affinity for nucleic acids. UNC4976 substantially potentiates the expression of polycomb target genes.
cell penetrant and potent positive allosteric modulator (PAM) of CBX7 chromodomain

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelsey N Lamb et al.
Cell chemical biology, 26(10), 1365-1379 (2019-08-20)
Polycomb-directed repression of gene expression is frequently misregulated in human diseases. A quantitative and target-specific cellular assay was utilized to discover the first potent positive allosteric modulator (PAM) peptidomimetic, UNC4976, of nucleic acid binding by CBX7, a chromodomain methyl-lysine reader

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service