Skip to Content
Merck
All Photos(1)

Key Documents

SML2908

Sigma-Aldrich

A71915 trifluoroacetate

≥95% (HPLC)

Synonym(s):

(Arg6, β-cyclohexyl-Ala8, D-Tic16, Arg17, Cys18)-Atrial Natriuretic Factor (6-18) amide (Cys7→Cys18 disulfide), TFA salt, A 71915, A-71915, Arg-Cys-β-cyclohexyl-Ala-Gly-Gly-Arg-Ile-Asp-Arg-Ile-D-Tic-Arg-Cys-NH2 (Cys2→Cys13 disulfide), TFA salt, RC-(Cha)-GGRIDRI-(D-Tic)-RC-NH2 (Cys2→Cys13 disulfide, TFA salt, [Arg6, Cha8]ANP-(6-15)-D-Tic-Arg-Cys-NH2 (Cys7→Cys18 disulfide), TFA salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C69H116N26O15S2 · xC2HF3O2
CAS Number:
Molecular Weight:
1613.95 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

A71915 is a potent atrial natriuretic peptide A (ANP, NPPA) receptor (NPR1, ANP-A, ANPR-A, NPR-A) antagonist (pKi = 9.18 (Ki = 650 nM) by competitive binding against 300 nM rat ANP1-28 to human neuroblastoma NB-OK-1 cells; pA2 = 9.45 against rat ANP-induced cGMP production in NB-OK-1 cells). A71915 is reactive toward dog, human, mouse, rabbit, and rat species, and commonly employed both in cultures (1-10 μM) and in vivo for studying NPR-A-mediated responses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuyuan Chu et al.
Peptides, 90, 1-9 (2017-02-24)
Atrial natriuretic peptide (ANP) is increasingly expressed on airway and inhibits pulmonary arterial remodeling. However, the role of ANP in remodeling of respiratory system is still unclear. The role of ANP on airway remodeling and the possible mechanism was explored
André M Leite-Moreira et al.
Cardiovascular research, 114(5), 656-667 (2018-02-06)
The heart is constantly challenged with acute bouts of stretching or overload. Systolic adaptations to these challenges are known but adaptations in diastolic stiffness remain unknown. We evaluated adaptations in myocardial stiffness due to acute stretching and characterized the underlying
G J Trachte
The Journal of pharmacology and experimental therapeutics, 264(3), 1227-1233 (1993-03-01)
Atrial natriuretic factor (ANF) suppresses adrenergic and purinergic neurotransmission in the rabbit vas deferens. The neuromodulatory mechanism of action for ANF is not established, but is thought to be independent of cyclic GMP (cGMP) generation. This study directly tested for
C Delporte et al.
European journal of pharmacology, 224(2-3), 183-188 (1992-12-02)
The effects of seven competitive atrial natriuretic peptide (ANP) receptor antagonists were compared on cultured human neuroblastoma NB-OK-1 cells expressing exclusively ANPA receptors, by evaluating their capacity to inhibit [125I]ANP binding and to suppress ANP-stimulated cyclic GMP elevation. In ANP
Kuichang Yuan et al.
Peptides, 82, 12-19 (2016-05-22)
The endothelins (ET) peptide family consists of ET-1, ET-2, ET-3, and sarafotoxin (s6C, a snake venom) and their actions appears to be different among isoforms. The aim of this study was to compare the secretagogue effect of ET-1 on atrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service