Skip to Content
Merck
All Photos(1)

Key Documents

SML2907

Sigma-Aldrich

Azumolene

≥98% (HPLC)

Synonym(s):

1-[[[5-(4-Bromophenyl)-2-oxazolyl]methylene]amino]-2,4-imidazolidinedione, EU 4093

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$146.00
25 MG
$593.00

$146.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$146.00
25 MG
$593.00

About This Item

Empirical Formula (Hill Notation):
C13H9BrN4O3
CAS Number:
Molecular Weight:
349.14
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$146.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

InChI

1S/C13H9BrN4O3/c14-9-3-1-8(2-4-9)10-5-15-12(21-10)6-16-18-7-11(19)17-13(18)20/h1-6H,7H2,(H,17,19,20)

InChI key

SEGCNGONCZQFDW-UHFFFAOYSA-N

Biochem/physiol Actions

Azumolene, a more water-soluble analog of dantrolene, is a potent inhibitor of sarcoplasmic reticulum (SR) ryanodine receptor (RyR) that inhibits skeletal muscle SR Ca2+ release. Azumolene is a direct acting skeletal muscle relaxant that induces effective blockade of skeletal muscle RyR1.
potent inhibitor of sarcoplasmic reticulum ryanodine receptor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin E Talbert et al.
PloS one, 11(2), e0148161-e0148161 (2016-02-06)
Mechanical ventilation (MV) is a life-saving intervention for patients in respiratory failure. However, prolonged MV causes the rapid development of diaphragm muscle atrophy, and diaphragmatic weakness may contribute to difficult weaning from MV. Therefore, developing a therapeutic countermeasure to protect
Daoyuan Si et al.
Journal of cardiovascular electrophysiology, 29(12), 1707-1715 (2018-09-12)
Following long-duration ventricular fibrillation (LDVF), reinitiation of ventricular fibrillation (VF) poses a major challenge during resuscitation. Ryanodine receptor 2 (RyR2) becomes dysfunctional following VF. The relationship between LDVF, RyR2 modulation, and ventricular refibrillation, as well as the role of RyR2 phosphorylation
Yingfan Zhang et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 94-102 (2005-04-16)
Azumolene is an analog of dantrolene, the only approved medicine for treatment of malignant hyperthermia (MH). The pharmacological mechanism of these drugs is to inhibit skeletal muscle sarcoplasmic reticulum (SR) Ca2+ release by modulating the activity of the SR ryanodine

Questions

  1. Do you have any clinical administration data in mailignant hyperthermia?

    1 answer
    1. This product is intended for laboratory research use only. Therefore, no clinical trial or other therapeutic information is available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service