Skip to Content
Merck
All Photos(1)

Key Documents

SML2888

Sigma-Aldrich

LH1306

≥95% (HPLC)

Synonym(s):

2,2’-Dimethyl-3,3’-bis[2-methoxy-3-(2-acetamidoethylamino)-methylpyridin-6-yl-oxymethyl]-1,1’-biphenyl, LH 1306, LH-1306, N,N′-(2,2′-(6,6′-(2,2′-Dimethylbiphenyl-3,3′-diyl)bis(methylene)bis(oxy)bis(2-methoxypyridine-6,3-diyl))bis(methylene)bis(azanediyl)bis(ethane-2,1-diyl))diacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$260.00
25 MG
$1,050.00

$260.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$260.00
25 MG
$1,050.00

About This Item

Empirical Formula (Hill Notation):
C38H48N6O6
CAS Number:
Molecular Weight:
684.82
UNSPSC Code:
12352200
NACRES:
NA.77

$260.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

-10 to -25°C

Biochem/physiol Actions

LH1306 is a potent programmed cell death-1/programmed death-ligand 1 (PD-1 (CD279, PDCD1)/PD-L1 (B7-H1, PDCD1-L1)) protein-protein interaction (PPI) inhibitor (IC50 = 25 nM by HTRF assay) that induces symmetrically arranged PD-L1 homodimer formation by targeting PD-L1 at the PD-1 binding site. LH1306 effectively blocks cell surface PD-1/PD-L1 interaction between PD-1-expressing Jurkat cells and co-cultured PD-L1-expressing U2OS cells (SHP-1 recruitment IC50 = 334 nM) or co-cultured PD-L1-expressing aAPC/CHO-K1 cells (NFAT signaling restoration EC50 = 4.21 μM).
Potent programmed cell death-1 (PD-1, CD279, PDCD1)/PD-1 ligand (PD-L1, B7-H1, PDCD1-L1) protein-protein interaction (PPI) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service