SML2881
R-VK4-40
≥98% (HPLC)
Synonym(s):
R-VK4-40, N-[(3R)-4-[4-(2-Chloro-3-ethylphenyl)-1-piperazinyl]-3-hydroxybutyl]-1H-indole-2-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C1=CC2=C(C=CC=C2)N1)NCC[C@H](CN3CCN(C4=CC=CC(CC)=C4Cl)CC3)O
Biochem/physiol Actions
R-VK4-40 is an orally available, highly selective and potent dopamine D3 receptor (D3R) antagonists that does not exhibit unwanted cardiovascular effects. R-VK4-40 attenuates oxycodone induced rewards without compromising antinociceptive effects in rats. It reduces oxycodone self-administration and suppress naloxone-caused opioid withdrawal.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Neuropharmacology, 158, 107597-107597 (2019-04-12)
Prescription opioid abuse is a global crisis. New treatment strategies for pain and opioid use disorders are urgently required. We evaluated the effects of R-VK4-40, a highly selective dopamine (DA) D3 receptor (D3R) antagonist, on the rewarding and analgesic effects
The Journal of pharmacology and experimental therapeutics, 371(3), 602-614 (2019-09-29)
Opioid and cocaine abuse are major public health burdens. Existing medications for opioid use disorder are limited by abuse liability and side effects, whereas no treatments are currently approved in the United States for cocaine use disorder. Dopamine D3 receptor
Journal of medicinal chemistry, 62(20), 9061-9077 (2019-09-19)
Dopamine D3 receptors (D3R) play a critical role in neuropsychiatric conditions including substance use disorders (SUD). Recently, we reported a series of N-(3-hydroxy-4-(4-phenylpiperazin-1-yl)butyl)-1H-indole-2-carboxamide analogues as high affinity and selective D3R lead molecules for the treatment of opioid use disorders (OUD).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service