Skip to Content
Merck
All Photos(1)

Key Documents

SML2812

Sigma-Aldrich

AN1

≥98% (HPLC)

Synonym(s):

5-Bromo-3-[(3-bromo-4,5-dihydroxyphenyl)methylene]-1,3-dihydro-2H-indol-2-one, mHTT-LC3 linker AN1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00
25 MG
$593.00

$168.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$168.00
25 MG
$593.00

About This Item

Empirical Formula (Hill Notation):
C15H9Br2NO3
CAS Number:
Molecular Weight:
411.04
UNSPSC Code:
12352200
NACRES:
NA.77

$168.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Brc1c(c(cc(c1)C=C2c3c(ccc(c3)Br)NC2=O)O)O

InChI

1S/C15H9Br2NO3/c16-8-1-2-12-9(6-8)10(15(21)18-12)3-7-4-11(17)14(20)13(19)5-7/h1-6,19-20H,(H,18,21)

InChI key

GPKLWRHHVIBYEO-UHFFFAOYSA-N

Biochem/physiol Actions

AN1 is a brain blood barrier penetrant agent that tether LC3 and mHTT and targets mHTT for autophagosome degradation. AN1 significantly lower mHTT in cortices of HD mice, in cells from patients with Huntington′s disease, and neurons derived from induced pluripotent stem cells. AN1 does not lower wtHTT in fibroblasts from healthy human donors. It appears that AN1 interact with the polyQ stretch.
brain blood barrier penetrant agent that tether LC3 and mHTT and targets mHTT for autophagosome degradation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoyang Li et al.
Nature, 575(7781), 203-209 (2019-11-02)
Accumulation of mutant proteins is a major cause of many diseases (collectively called proteopathies), and lowering the level of these proteins can be useful for treatment of these diseases. We hypothesized that compounds that interact with both the autophagosome protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service