Skip to Content
Merck
All Photos(1)

Key Documents

SML2797

Sigma-Aldrich

UNBS5162

≥98% (HPLC)

Synonym(s):

N-[2-[2-(Dimethylamino)ethyl]-2,3-dihydro-1,3-dioxo-1H-benz[de]isoquinolin-5-yl]urea, UNBS 5162, [2-[2-(Dimethylamino)ethyl]-1,3-dioxobenzo[de]isoquinolin-5-yl]urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$156.00
25 MG
$575.00

$156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$156.00
25 MG
$575.00

About This Item

Empirical Formula (Hill Notation):
C17H18N4O3
CAS Number:
Molecular Weight:
326.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$156.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]1([c](c2c3c([c]1=O)cccc3cc(c2)NC(=O)N)=O)CCN(C)C

InChI

1S/C17H18N4O3/c1-20(2)6-7-21-15(22)12-5-3-4-10-8-11(19-17(18)24)9-13(14(10)12)16(21)23/h3-5,8-9H,6-7H2,1-2H3,(H3,18,19,24)

InChI key

WCKZRLOUKYFJDY-UHFFFAOYSA-N

Biochem/physiol Actions

UNBS5162, an amonafide (naphthalimide) derivative, is a potent antitumor agent that inhibits tumor proliferation, progression and invasiveness. UNBS5162 is a pan-antagonist of CXCL chemokine expression. It appears that UNBS5162 inhibits melanoma progression through inhibition of the AKT/mTOR signaling pathway.
pan-antagonist of CXCL chemokine expression

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatjana Mijatovic et al.
Neoplasia (New York, N.Y.), 10(6), 573-586 (2008-06-03)
Several naphthalimides have been evaluated clinically as potential anticancer agents. UNBS3157, a naphthalimide that belongs to the same class as amonafide, was designed to avoid the specific activating metabolism that induces amonafide's hematotoxicity. The current study shows that UNBS3157 rapidly
Xin Yue et al.
Experimental and therapeutic medicine, 16(5), 3921-3928 (2018-10-23)
UNBS5162, a naphthalimide derivative, has a strong anticancer effect and low clinical hematological toxicity. UNBS5162 has been shown to have an anticancer effect in numerous cancer types, except triple negative breast cancer (TNBC). To explore this issue, TNBC MDA-MB-231 cells
Yingyi Ye et al.
Cancer management and research, 11, 2339-2348 (2019-04-10)
Human melanoma is a malignant tumor originated from melanocytes with high invasion, metastasis, and poor prognosis. In this study, the effects of naphthalimides UNBS5162 and amonafide on the properties of proliferation and apoptosis in human melanoma cells were confirmed. Cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service