Skip to Content
Merck
All Photos(1)

Key Documents

SML2790

Sigma-Aldrich

YU254403

≥98% (HPLC)

Synonym(s):

4-{7,9-dichloro-1H,2H,3H,3aH,4H,5H,9bH-cyclopenta[c]quinolin-4-yl}benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$134.00
25 MG
$545.00

$134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$134.00
25 MG
$545.00

About This Item

Empirical Formula (Hill Notation):
C19H17Cl2NO2
CAS Number:
Molecular Weight:
362.25
UNSPSC Code:
51111800
NACRES:
NA.77

$134.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C(NC2=CC(Cl)=C3)C4CCCC4C2=C3Cl)C=C1)O

InChI

1S/C19H17Cl2NO2/c20-12-8-15(21)17-13-2-1-3-14(13)18(22-16(17)9-12)10-4-6-11(7-5-10)19(23)24/h4-9,13-14,18,22H,1-3H2,(H,23,24)

InChI key

PHDZTYHPMHMZPG-UHFFFAOYSA-N

Biochem/physiol Actions

YU254403 is a potent inhibitor of phosphatidylserine decarboxylases (PSD) including Plasmodium knowlesi phosphatidylserine decarboxylases (PkPSD). YU254403 potently inhibits growth of pathogenic yeast Candida albicans, and other pathogens including Aspergillus fumigatus and Fusarium solani.
potent inhibitor of phosphatidylserine decarboxylases (PSD)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Hendricson et al.
The Journal of biological chemistry, 294(32), 12146-12156 (2019-06-23)
Phosphatidylserine decarboxylases (PSDs) catalyze the decarboxylation of phosphatidylserine to generate phosphatidylethanolamine, a critical step in phospholipid metabolism in both prokaryotes and eukaryotes. Most PSDs are membrane-bound, and classical radioisotope-based assays for determining their activity in vitro are not suitable for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service