Skip to Content
Merck
All Photos(1)

Key Documents

SML2789

Sigma-Aldrich

tBPC

≥98% (HPLC)

Synonym(s):

2-[4-(1,1-Dimethylethyl)phenoxy]cyclohexanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$83.40
25 MG
$340.00

$83.40


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$83.40
25 MG
$340.00

About This Item

Empirical Formula (Hill Notation):
C16H24O2
CAS Number:
Molecular Weight:
248.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$83.40


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O(C2C(CCCC2)O)c1ccc(cc1)C(C)(C)C

InChI

1S/C16H24O2/c1-16(2,3)12-8-10-13(11-9-12)18-15-7-5-4-6-14(15)17/h8-11,14-15,17H,4-7H2,1-3H3

InChI key

FTIXUILRMBSXNS-UHFFFAOYSA-N

Biochem/physiol Actions

Postitive allosteric modulator of NPY Y4 receptor
tBPC is a postitive allosteric modulator of the neuropeptide Y (NPY) Y4 receptor, which is predominantly expressed in the gastrointestinal tract and activated most strongly by pancreatic polypeptide (PP). tBPC was shown to potentiate Y4R activity in G-Protein signaling and arrestin3 recruitment and to potentiate the native PP response. tBPC was selective for Y4R over other Neuropeptide Y receptors and showed no effect on the efficacy (Emax) or potency (EC50) of the NPY signal response at the Y1, Y2, or Y5 receptors at 30 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Schubert et al.
Journal of medicinal chemistry, 60(17), 7605-7612 (2017-08-11)
The human Y4 receptor (Y4R) and its cognate ligand, pancreatic polypeptide (PP), are involved in the regulation of energy expenditure, satiety, and food intake. This system represents a potential target for the treatment of metabolic diseases and has been extensively

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service