Skip to Content
Merck
All Photos(1)

Key Documents

SML2780

Sigma-Aldrich

ML402

≥98% (HPLC)

Synonym(s):

N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-thiophenecarboxamide, N-[2-(4-Chloro-2-methylphenoxy)ethyl]thiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$627.00

$141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$627.00

About This Item

Empirical Formula (Hill Notation):
C14H14ClNO2S
CAS Number:
Molecular Weight:
295.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$141.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC=C(OCCNC(C2=CC=CS2)=O)C(C)=C1

InChI

1S/C14H14ClNO2S/c1-10-9-11(15)4-5-12(10)18-7-6-16-14(17)13-3-2-8-19-13/h2-5,8-9H,6-7H2,1H3,(H,16,17)

InChI key

RULQUKFOBAPKKR-UHFFFAOYSA-N

Biochem/physiol Actions

Activator of K2P2.1 (TREK-1) and K2P10.1 (TREK-2) channels
ML402 is a selective activator of K2P2.1 (TREK-1) and K2P10.1 (TREK-2) potassium channels. It was used to determine a cryptic selectivity filter binding site of the K2P2.1 (TREK-1) channel, designated the K2P modulator pocket. This binding pocket stabilizes an active "leak-mode" conformation, binding small-molecule activators that restrict the mobility of the selectivity filter and surrounding structure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Lolicato et al.
Nature, 547(7663), 364-368 (2017-07-12)
Polymodal thermo- and mechanosensitive two-pore domain potassium (K2P) channels of the TREK subfamily generate 'leak' currents that regulate neuronal excitability, respond to lipids, temperature and mechanical stretch, and influence pain, temperature perception and anaesthetic responses. These dimeric voltage-gated ion channel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service