Skip to Content
Merck
All Photos(1)

Key Documents

SML2774

Sigma-Aldrich

KDOAM25 hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

2-[[[2-[2-(Dimethylamino)ethyl-ethyl-amino]-2-oxidanylidene-ethyl]amino]methyl]pyridine-4-carboxamide hydrochloride hydrate, 2-[[[2-[2-(Dimethylamino)ethyl-ethylamino]-2-oxoethyl]amino]methyl]pyridine-4-carboxamide hydrochloride hydrate, KDOAM-25 hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$174.00
25 MG
$665.00

$174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$174.00
25 MG
$665.00

About This Item

Empirical Formula (Hill Notation):
C15H25N5O2 · xHCl · yH2O
Molecular Weight:
307.39 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$174.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N(CCN(CC)C(=O)CNCc1nccc(c1)C(=O)N)(C)C

InChI key

PNFMVADNCOGWME-UHFFFAOYSA-N

Biochem/physiol Actions

KDOAM25 is a selective inihbitor of the KDM5 family histone demethylases JARID1A, JARID1B, JARID1C and JARID1D with IC50 values < 60 nM. JARID1A and JARID1B are independently overexpressed in some cancers with JARID1B (KDM5B, PLU1) also identified as a potential oncogene, a repressor of tumour repressor genes. JARID1C (KDM5C) and JARID1D (KDM5D) are located on the X- and Y-chromosomes respectively. KDOAM25′s closest off-target is JMJD2C (selectivity >400 fold). It shows no activity on other tested 2-OG family members, including FIH, NO66, MINA53 and PHD2. KDOAM25 is active in cells.
Selective inihbitor of the KDM5 family histone demethylases JARID1A, JARID1B, JARID1C and JARID1D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica M Montano et al.
Breast cancer research : BCR, 21(1), 138-138 (2019-12-07)
The tumor suppressor actions of hexamethylene bis-acetamide (HMBA)-inducible protein 1 (HEXIM1) in the breast, prostate, melanomas, and AML have been reported by our group and others. Increased HEXIM1 expression caused differentiation and inhibited proliferation and metastasis of cancer cells. Historically
Anthony Tumber et al.
Cell chemical biology, 24(3), 371-380 (2017-03-07)
Methylation of lysine residues on histone tail is a dynamic epigenetic modification that plays a key role in chromatin structure and gene regulation. Members of the KDM5 (also known as JARID1) sub-family are 2-oxoglutarate (2-OG) and Fe2+-dependent oxygenases acting as
Simone Pippa et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-08)
Background: KDM5 enzymes are H3K4 specific histone demethylases involved in transcriptional regulation and DNA repair. These proteins are overexpressed in different kinds of cancer, including breast, prostate and bladder carcinomas, with positive effects on cancer proliferation and chemoresistance. For these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service