Skip to Content
Merck
All Photos(1)

Key Documents

SML2736

Sigma-Aldrich

NPBA

≥98% (HPLC)

Synonym(s):

N-(2-((4-nitro-2-(trifluoromethyl)phenyl)amino)ethyl)benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$154.00
25 MG
$566.00

$154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$154.00
25 MG
$566.00

About This Item

Empirical Formula (Hill Notation):
C16H14F3N3O3
CAS Number:
Molecular Weight:
353.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$154.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(ccc(c1)[N+](=O)[O-])NCCNC(=O)c2ccccc2

InChI key

LUJDPMMSYZQCHA-UHFFFAOYSA-N

Biochem/physiol Actions

NPBA is a potent and selective agonist (activator) of potassium K2P channel TASK-3 (KCNK9).
potent and selective agonist (activator) of potassium K2P channel TASK-3 (KCNK9)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuyun Tian et al.
Molecular pharmacology, 96(1), 26-35 (2019-04-25)
The TASK-3 channel is a member of the K2P family that is important for the maintenance of the resting membrane potential. Previous studies have demonstrated that the TASK-3 channel is involved in several physiologic and pathologic processes, including sleep/wake control

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service