Skip to Content
Merck
All Photos(1)

Key Documents

SML2721

Sigma-Aldrich

GSK8612

≥98% (HPLC)

Synonym(s):

4-(((5-Bromo-2-((3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-yl)amino)pyrimidin-4-yl)amino)methyl)benzenesulfonamide, 4-((5-Bromo-2-(3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-ylamino)pyrimidin-4-ylamino)methyl)benzenesulfonamide, GSK 8612, GSK-8612

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$269.00
25 MG
$889.00

$269.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$269.00
25 MG
$889.00

About This Item

Empirical Formula (Hill Notation):
C17H17BrF3N7O2S
CAS Number:
Molecular Weight:
520.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$269.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C[n]1nc(c(c1)Nc2nc(c(cn2)Br)NCc3ccc(cc3)[S](=O)(=O)N)C

InChI key

FFPHMUIGESPOTK-UHFFFAOYSA-N

Biochem/physiol Actions

GSK8612 is a potent and highly selective tank-binding kinase-1 (TBK1) inhibitor (pIC50 = 6.8; pKd = 8.0 vs pKa = 6.2/STK17B, 6.0/IKKε, 6.1/LRRK2, 6.0/FLT4, 5.8/ULK3, 5.7/BMP2K, 5.7/CSNK2A1, 5.6/CSNK2A2, 5.6/FLT4, 5.5/PIK3CD, <5.5/more than 270 kinases). GSK8612 inhibits IFNβ secretion induced by dsDNA containing virus or STING ligand cGAMP (IC50 = 1.25 & 0.5 μM, respectively; THP-1), as well as suppresses TLR3 ligand poly(I:C)-induced IRF3 Ser396 phosphorylation (IC50 = 1 μM; Ramos) and IFNα release (IC50 = 0.79 μM; human PBMCs), a IFN positive feedback response steming from IFNβ.
Potent and highly selective tank-binding kinase-1 (TBK1) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas W Thomson et al.
ACS medicinal chemistry letters, 10(5), 780-785 (2019-05-18)
The serine/threonine protein kinase TBK1 (Tank-binding Kinase-1) is a noncanonical member of the IkB kinase (IKK) family. This kinase regulates signaling pathways in innate immunity, oncogenesis, energy homeostasis, autophagy, and neuroinflammation. Herein, we report the discovery and characterization of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service