Skip to Content
Merck
All Photos(1)

Key Documents

SML2704

Sigma-Aldrich

BAY-294

≥98% (HPLC)

Synonym(s):

BAY-294, 6,7-Dimethoxy-2-methyl-N-[(S)-1-(4-{2-[(methylamino)methyl]phenyl}thien-2-yl)ethyl]quinazolin-4-amine, BAY 293 (S)-enantiomer, BAY 293 control, BAY 294, BAY-293 (S)-enantiomer, BAY-293 control, BAY293 (S)-enantiomer, BAY293 control, BAY294

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$176.00
25 MG
$705.00

$176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$176.00
25 MG
$705.00

About This Item

Empirical Formula (Hill Notation):
C25H28N4O2S
CAS Number:
Molecular Weight:
448.58
UNSPSC Code:
12352200
NACRES:
NA.77

$176.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +86.0 to +106.0°, c = 1.0 in DMSO

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@@H](C1=CC(C2=CC=CC=C2CNC)=CS1)NC3=C4C=C(C(OC)=CC4=NC(C)=N3)OC

InChI

1S/C25H28N4O2S/c1-15(24-10-18(14-32-24)19-9-7-6-8-17(19)13-26-3)27-25-20-11-22(30-4)23(31-5)12-21(20)28-16(2)29-25/h6-12,14-15,26H,13H2,1-5H3,(H,27,28,29)/t15-/m0/s1

InChI key

WEGLOYDTDILXDA-HNNXBMFYSA-N

Biochem/physiol Actions

BAY-294 corresponds to the (S)-enantiomer of and the recommended control for the selective SOS1-KRAS interaction inhibitor BAY-293 (IC50 = 2.34 μM/BAY-294 vs. 21 nM/BAY-293; 10 nM hSOS1cat & 5 nM hKRAS G12C). BAY-294 exhibits little potency against SOS1-mediated KRAS activation (IC50 = 3.91 μM/BAY-294 vs. 52 nM/BAY-293; 10 nM SOS1cat & 50 nM wt hKRAS).
Control (S)-enantiomer of the selective SOS1-KRAS interaction inhibitor BAY-293.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service