Skip to Content
Merck
All Photos(1)

Key Documents

SML2682

Sigma-Aldrich

RIPGBM

≥98% (HPLC)

Synonym(s):

N-(3-(Benzylamino)-1,4-dioxo-1,4-dihydronaphthalen-2-yl)-N-(4-fluorobenzyl)acetamide, N-[1,4-Dihydro-1,4-dioxo-3-[(phenylmethyl)amino]-2-naphthalenyl]-N-[(4-fluorophenyl)methyl]acetamide, cRIPGBM precursor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$134.00
25 MG
$538.00

$134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$134.00
25 MG
$538.00

About This Item

Empirical Formula (Hill Notation):
C26H21FN2O3
CAS Number:
Molecular Weight:
428.45
UNSPSC Code:
12352200
NACRES:
NA.77

$134.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

faint brown to very dark brown-red

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(N(CC2=CC=C(F)C=C2)C(C)=O)=C(NCC3=CC=CC=C3)C(C4=C1C=CC=C4)=O

InChI

1S/C26H21FN2O3/c1-17(30)29(16-19-11-13-20(27)14-12-19)24-23(28-15-18-7-3-2-4-8-18)25(31)21-9-5-6-10-22(21)26(24)32/h2-14,28H,15-16H2,1H3

InChI key

COATXBHZYVUJQP-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, brain-penetrant, GBM CSCs-selective apoptosis inducer with efficacy in a murine orthotopic xenograft model of glioblastoma multiforme in vivo.
RIPGBM is a brain-penetrant, orally available, glioblastoma multiforme (GBM) cancer stem cells (CSCs)-selective apoptosis inducer (GBM-1/-5/-39 EC50 = 220/290/890 nM; human neural progenitor cells (NPCs)/astrocytes/lung fibroblasts EC50 = 1.7/2.8/3.5 μM) with in vivo efficacy against intracranial human GBM orthotopic xenograft tumor growth in mice (50 mg/kg bid. po.). RIPGBM undergoes a cell type-selective metabolic redox conversion to preferentially form in GBM CSCs the derivative cRIPGBM that binds receptor-interacting protein kinase 2 (RIPK2, RIP2) and promotes the formation of a proapoptotic RIPK2/caspase-1 complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha C Lucki et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(13), 6435-6440 (2019-03-09)
Glioblastoma multiforme (GBM; grade IV astrocytoma) is the most prevalent and aggressive form of primary brain cancer. A subpopulation of multipotent cells termed GBM cancer stem cells (CSCs) play a critical role in tumor initiation, tumor maintenance, metastasis, drug resistance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service