Skip to Content
Merck
All Photos(1)

Key Documents

SML2679

Sigma-Aldrich

PFF-1

≥97% (HPLC)

Synonym(s):

3′,6′-Bis(diethylamino)-2-[(E)-(2-pyridinylmethylene)amino]-spiro[1H-isoindole-1,9′-[9H]xanthen]-3(2H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$131.00
25 MG
$530.00

$131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$131.00
25 MG
$530.00

About This Item

Empirical Formula (Hill Notation):
C34H35N5O2
CAS Number:
Molecular Weight:
545.67
UNSPSC Code:
12352200
NACRES:
NA.77

$131.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N2(C4(c5c(cc(cc5)N(CC)CC)Oc6c4ccc(c6)N(CC)CC)c3c(cccc3)C2=O)\N=C\c1ncccc1

InChI key

APANOTMHLDBMQU-GOJREDGKSA-N

Biochem/physiol Actions

PFF-1 is a robust, cell penetrant and long time-laps (>30 min) fluorophore suitable for single-molecule localization microscopy experiments (SMLM). PFF-1 exhibits minimal phototoxicity and no apparent photobleaching. It was used to visualize movement of vesicles within cultured neurons.
robust, cell penetrant and long time-laps (>30 min) fluorophore

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward a highly sensitive and selective indole-rhodamine-based light-up probe for Hg2+ and its application in living cells
Yao T, Guo-Fang J
Tetrahedron Letters, 58, 2846-2849 null
Elias A Halabi et al.
Nature communications, 10(1), 1232-1232 (2019-03-16)
Photoswitchable molecules have multiple applications in the physical and life sciences because their properties can be modulated with light. Fluxional molecules, which undergo rapid degenerate rearrangements in the electronic ground state, also exhibit switching behavior. The stochastic nature of fluxional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service