Skip to Content
Merck
All Photos(1)

Key Documents

SML2666

Sigma-Aldrich

Diflapolin

≥98% (HPLC)

Synonym(s):

N-[4-(2-Benzothiazolylmethoxy)-2-methylphenyl]-N′-(3,4-dichlorophenyl)-urea, N-[4-(Benzothiazol-2-ylmethoxy)-2-methylphenyl]-N′-(3,4-dichlorophenyl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$139.00
25 MG
$563.00

$139.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$139.00
25 MG
$563.00

About This Item

Empirical Formula (Hill Notation):
C22H17Cl2N3O2S
CAS Number:
Molecular Weight:
458.36
UNSPSC Code:
41116004
NACRES:
NA.77

$139.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(Cl)C(Cl)=C1)NC2=C(C)C=C(OCC3=NC4=CC=CC=C4S3)C=C2

InChI

1S/C22H17Cl2N3O2S/c1-13-10-15(29-12-21-26-19-4-2-3-5-20(19)30-21)7-9-18(13)27-22(28)25-14-6-8-16(23)17(24)11-14/h2-11H,12H2,1H3,(H2,25,27,28)

InChI key

FGXLEECGXSDIMM-UHFFFAOYSA-N

Biochem/physiol Actions

Diflapolin is a bioavailable, highly potent and highly selective dual FLAP/sEH (5-LO-activating protein/ soluble epoxide hydrolase) inhibitor that suppresses leukotriene formation and increases epoxyeicosatrienoic acids levels. Diflapolin exhibit potent anti-inflammatory activities in mice models.
bioavailable, highly potent and highly selective dual FLAP/sEH (5-LO-activating protein/ soluble epoxide hydrolase) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Vieider et al.
ACS medicinal chemistry letters, 10(1), 62-66 (2019-01-19)
A series of derivatives of the potent dual soluble epoxide hydrolase (sEH)/5-lipoxygenase-activating protein (FLAP) inhibitor diflapolin was designed, synthesized, and characterized by 1H NMR, 13C NMR, and elemental analysis. These novel compounds were biologically evaluated for their inhibitory activity against
Veronika Temml et al.
Scientific reports, 7, 42751-42751 (2017-02-22)
Leukotrienes (LTs) are pro-inflammatory lipid mediators derived from arachidonic acid (AA) with roles in inflammatory and allergic diseases. The biosynthesis of LTs is initiated by transfer of AA via the 5-lipoxygenase-activating protein (FLAP) to 5-lipoxygenase (5-LO). FLAP inhibition abolishes LT
Ulrike Garscha et al.
Scientific reports, 7(1), 9398-9398 (2017-08-26)
Arachidonic acid (AA) is metabolized to diverse bioactive lipid mediators. Whereas the 5-lipoxygenase-activating protein (FLAP) facilitates AA conversion by 5-lipoxygenase (5-LOX) to pro-inflammatory leukotrienes (LTs), the soluble epoxide hydrolase (sEH) degrades anti-inflammatory epoxyeicosatrienoic acids (EETs). Accordingly, dual FLAP/sEH inhibition might

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service