Skip to Content
Merck
All Photos(1)

Key Documents

SML2658

Sigma-Aldrich

TT-10

≥98% (HPLC)

Synonym(s):

N2-Allyl-5-[(5-fluorothiophen-2-yl)carbonyl]thiazole-2,4-diamine, TAZ K, TAZ-K, TT 10, [4-Amino-2-(2-propen-1-ylamino)-5-thiazolyl](5-fluoro-2-thienyl)methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$770.00

$166.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$770.00

About This Item

Empirical Formula (Hill Notation):
C11H10FN3OS2
CAS Number:
Molecular Weight:
283.35
UNSPSC Code:
41106609
NACRES:
NA.77

$166.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(C(C2=CC=C(S2)F)=O)SC(NCC=C)=N1

InChI

1S/C11H10FN3OS2/c1-2-5-14-11-15-10(13)9(18-11)8(16)6-3-4-7(12)17-6/h2-4H,1,5,13H2,(H,14,15)

InChI key

GVYPHJQPOHDZEI-UHFFFAOYSA-N

Biochem/physiol Actions

TAZ activator that promotes cardiomyocytes proliferation by activating YAP/TEAD-mediated transcription, as well as by exerting antioxidant and antiapoptotic activity.
TT-10 (TAZ K) is a TAZ (transcriptional coactivator with PDZ-binding motif) activator that promotes cardiomyocytes (CMs) proliferation (fold-increase by 10 μM TT-10 over untreated control = 5.2/rat CMs/40 hrs & 2.6/human iPSC-derived CMs/48 hrs) by activating YAP (YES-associated protein)/TEAD (transcriptional enhancer factor domain) complex-mediated transcription, as well as by exerting antioxidant and antiapoptotic activity. TT-10 ameliorates myocardial infarction-induced cardiac dysfunction in mice in vivo (10 mg/kg i.p.) at least in part via enhancing clonal expansion of existing cardiomyocytes with nuclear YAP expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hironori Hara et al.
JACC. Basic to translational science, 3(5), 639-653 (2018-11-21)
Accumulating data suggest that new cardiomyocytes in adults are generated from existing cardiomyocytes throughout life. To enhance the endogenous cardiac regeneration, we performed chemical screenings to identify compounds that activate pro-proliferative YES-associated protein and transcriptional enhancer factor domain activities in
Masamichi Ito et al.
Journal of molecular and cellular cardiology, 128, 90-95 (2019-01-27)
Since regenerative capacity of adult mammalian myocardium is limited, activation of the endogenous proliferative capacity of existing cardiomyocytes is a potential therapeutic strategy for treating heart diseases accompanied by cardiomyocyte loss. Recently, we performed a compound screening and developed a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service