Skip to Content
Merck
All Photos(1)

Key Documents

SML2636

Sigma-Aldrich

Sb4

≥98% (HPLC)

Synonym(s):

2-[[(4-Bromophenyl)methyl]thio]benzoxazole, BMP signaling agonist Sb4

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$164.00
25 MG
$528.00

$164.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$164.00
25 MG
$528.00

About This Item

Empirical Formula (Hill Notation):
C14H10BrNOS
CAS Number:
Molecular Weight:
320.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$164.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=CC=C(CSC2=NC3=C(C=CC=C3)O2)C=C1

InChI

1S/C14H10BrNOS/c15-11-7-5-10(6-8-11)9-18-14-16-12-3-1-2-4-13(12)17-14/h1-8H,9H2

InChI key

JDVOIBFEFVKUPL-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, potent bone morphogenetic protein (BMP) signaling agonist that rapidly activate BMP in human renal cells (HEK293)
Sb4 is a cell penetrant, potent bone morphogenetic protein (BMP) signaling agonist that rapidly activate BMP in human renal cells (HEK293). Sb4 activates BMP by stabilization of intracellular p-SMAD-1/5/9. Apparently activation of the BMP pathway by sb4 cannot be reversed by inhibition of Noggin or type I BMP kinase inhibitors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shayna T J Bradford et al.
The Journal of biological chemistry, 294(9), 3125-3136 (2019-01-04)
Bone morphogenetic protein (BMP) signaling is critical in renal development and disease. In animal models of chronic kidney disease (CKD), re-activation of BMP signaling is reported to be protective by promoting renal repair and regeneration. Clinical use of recombinant BMPs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service