Skip to Content
Merck
All Photos(1)

Key Documents

SML2539

Sigma-Aldrich

NAB-14

≥98% (HPLC)

Synonym(s):

Carbamic acid, N,N-diethyl-, 4-[(1H-indol-7-ylamino)carbonyl]phenyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$125.00
25 MG
$629.00

$125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$125.00
25 MG
$629.00

About This Item

Empirical Formula (Hill Notation):
C20H21N3O3
CAS Number:
Molecular Weight:
351.40
UNSPSC Code:
12352200
NACRES:
NA.77

$125.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)C(OC(C=C1)=CC=C1C(NC2=C3C(C=CN3)=CC=C2)=O)=O

InChI

1S/C20H21N3O3/c1-3-23(4-2)20(25)26-16-10-8-15(9-11-16)19(24)22-17-7-5-6-14-12-13-21-18(14)17/h5-13,21H,3-4H2,1-2H3,(H,22,24)

InChI key

WCGIKJOMEYPSIP-UHFFFAOYSA-N

Biochem/physiol Actions

NAB-14 is a negative allosteric modulator (NAM) selective for GluN2C/2D-containing NMDA glutamate receptors. NAB-14 was >800-fold selective for recombinant GluN2C/GluN2D over GluN2A/GluN2B in Xenopus oocytes and exhibited an IC50 value of 580 nM at recombinant GluN1/GluN2D receptors.
Negative allosteric modulator selective for GluN2C/2D-containing NMDA receptors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon A Swanger et al.
ACS chemical neuroscience, 9(2), 306-319 (2017-10-19)
N-Methyl-d-aspartate receptors (NMDARs) are ionotropic glutamate receptors that mediate excitatory synaptic transmission and have been implicated in numerous neurological disorders. NMDARs typically comprise two GluN1 and two GluN2 subunits. The four GluN2 subtypes (GluN2A-GluN2D) have distinct functional properties and gene
Lulu Yao et al.
Nature communications, 9(1), 4000-4000 (2018-10-03)
Selective disruption of synaptic drive to inhibitory neurons could contribute to the pathophysiology of various brain disorders. We have previously identified a GluN2A-selective positive allosteric modulator, GNE-8324, that selectively enhances N-methyl-D-aspartate receptor (NMDAR)-mediated synaptic responses in inhibitory but not excitatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service