Skip to Content
Merck
All Photos(1)

Key Documents

SML2537

Sigma-Aldrich

NSAH

≥98% (HPLC)

Synonym(s):

2-Hydroxy-benzoic acid (2E)-2-[(2-hydroxy-1-naphthalenyl)methylene]hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$113.00
25 MG
$363.00

$113.00


Estimated to ship on12 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$113.00
25 MG
$363.00

About This Item

Empirical Formula (Hill Notation):
C18H14N2O3
CAS Number:
Molecular Weight:
306.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$113.00


Estimated to ship on12 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=CC=C1C(N/N=C/C2=C(C=CC=C3)C3=CC=C2O)=O

InChI

1S/C18H14N2O3/c21-16-8-4-3-7-14(16)18(23)20-19-11-15-13-6-2-1-5-12(13)9-10-17(15)22/h1-11,21-22H,(H,20,23)/b19-11+

InChI key

ZZSJOLDICPVVAV-YBFXNURJSA-N

Biochem/physiol Actions

NSAH is a cell penetrant, potent and selective nonnucleoside reversible inhibitor of large subunit (hRRM1) of hRR (human ribonucleotide reductase). NSAH binds to catalytic site of hRRM1. It inhibits hRR in cells. NSAH exhibit potent cytotoxicity toward cancer cells, while demonstrated very moderate toxicity against normal blood progenitor cells.
cell penetrant, potent and selective nonnucleoside reversible inhibitor of large subunit (hRRM1) of hRR (human ribonucleotide reductase)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Faiz Ahmad et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(31), 8241-8246 (2017-07-19)
Human ribonucleotide reductase (hRR) is crucial for DNA replication and maintenance of a balanced dNTP pool, and is an established cancer target. Nucleoside analogs such as gemcitabine diphosphate and clofarabine nucleotides target the large subunit (hRRM1) of hRR. These drugs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service