Skip to Content
Merck
All Photos(1)

Key Documents

SML2522

Sigma-Aldrich

Diprovocim-1

≥98% (HPLC)

Synonym(s):

(3S,3′S,4S,4′S)-1,1′-(1,4-Phenylenedicarbonyl)bis[N-[(1S,2R)-2-phenylcyclopropyl]-3,4-pyrrolidinedicarboxamide, 3,4-Pyrrolidinedicarboxamide, 1,1′-(1,4-phenylenedicarbonyl)bis[N-[(1S,2R)-2-phenylcyclopropyl]-, (3S,3′S,4S,4′S)-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$269.00
25 MG
$1,090.00

$269.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$269.00
25 MG
$1,090.00

About This Item

Empirical Formula (Hill Notation):
C56H56N6O6
CAS Number:
Molecular Weight:
909.08
UNSPSC Code:
51111800
NACRES:
NA.77

$269.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N7(C[C@H]([C@@H](C7)C(=O)N[C@@H]%10[C@H](C%10)c%11ccccc%11)C(=O)N[C@@H]8[C@H](C8)c9ccccc9)C(=O)c1ccc(cc1)C(=O)N2C[C@H]([C@@H](C2)C(=O)N[C@@H]5[C@H](C5)c6ccccc6)C(=O)N[C@@H]3[C@H](C3)c4ccccc4

InChI key

ABZBNXFGYUSVCJ-UYMKNZQYSA-N

Biochem/physiol Actions

Agonist of toll-like receptors TLR2 and TLR1
Diprovocim-1 is a potent agonist of toll-like receptors TLR2 and TLR1 with an EC50 value of 100 pM. Diprovocim-1 was shown to synergize with anti-PD-L1 treatment, inhibiting tumor growth, generating long-term antitumor memory, and curing or prolonging survival of mice engrafted with the murine melanoma B16-OVA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(37), E8698-E8706 (2018-08-29)
Successful cancer immunotherapy entails activation of innate immune receptors to promote dendritic cell (DC) maturation, antigen presentation, up-regulation of costimulatory molecules, and cytokine secretion, leading to activation of tumor antigen-specific cytotoxic T lymphocytes (CTLs). Here we screened a synthetic library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service