Skip to Content
Merck
All Photos(1)

Key Documents

SML2460

Sigma-Aldrich

SK33

≥98% (HPLC)

Synonym(s):

3-[3,5-Bis(trifluoromethyl)phenoxy]-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$133.00
25 MG
$428.00

$133.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$133.00
25 MG
$428.00

About This Item

Empirical Formula (Hill Notation):
C20H13F9N2O3
CAS Number:
Molecular Weight:
500.31
UNSPSC Code:
12352200
NACRES:
NA.77

$133.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C(NC1=CC(C(F)(F)F)=C(C#N)C=C1)=O)(C)COC2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2

InChI

1S/C20H13F9N2O3/c1-17(33,16(32)31-13-3-2-10(8-30)15(7-13)20(27,28)29)9-34-14-5-11(18(21,22)23)4-12(6-14)19(24,25)26/h2-7,33H,9H2,1H3,(H,31,32)

InChI key

VJIVKRQGSGBLFP-UHFFFAOYSA-N

Biochem/physiol Actions

SK33, a trifluoromethylated enobosarm analog, is a cell and brain penetrant, tissue selective and highly potent anti-androgen that reduces androgen receptor (AR) transcriptional activity. SK33 induces cell cycle arrest at G1 phase. It exhibits increased efficacy against acquired anti-androgen resistance prostate cancer cells.
cell and brain penetrant, tissue selective and highly potent anti-androgen that reduces androgen receptor (AR) transcriptional activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Alwyn Dart et al.
Molecular cancer therapeutics, 17(9), 1846-1858 (2018-06-14)
Prostate cancer often develops antiandrogen resistance, possibly via androgen receptor (AR) mutations, which change antagonists to agonists. Novel therapies with increased anticancer activity, while overcoming current drug resistance are urgently needed. Enobosarm has anabolic effects on muscle and bone while
Salvatore Ferla et al.
Bioorganic & medicinal chemistry letters, 26(15), 3636-3640 (2016-06-16)
Prostate cancer is a major cause of male death worldwide and the identification of new and improved treatments is constantly required. Among the available options, different non-steroidal androgen receptor (AR) antagonists are approved also to treat castration-resistant forms. Most of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service