Skip to Content
Merck
All Photos(1)

Key Documents

SML2422

Sigma-Aldrich

Sophocarpine

≥95% (HPLC)

Synonym(s):

(-)-Sophocarpine, 13,14-Didehydromatridin-15-one, SC

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$69.60
125 MG
$212.00

$69.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$69.60
125 MG
$212.00

About This Item

Empirical Formula (Hill Notation):
C15H22N2O
CAS Number:
Molecular Weight:
246.35
MDL number:
UNSPSC Code:
12352200

$69.60


Check Cart for Availability

Request a Bulk Order

biological source

Sophora alopecuroides

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D -26 to -35°, c = 0.5 in ethanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[Br-].N21[C@@H]3[C@@H]([C@@H]4N(C[C@@H]3CCC2)C(=O)C=CC4)CCC1.[H+]

InChI

1S/C15H22N2O.BrH/c18-14-7-1-6-13-12-5-3-9-16-8-2-4-11(15(12)16)10-17(13)14;/h1,7,11-13,15H,2-6,8-10H2;1H/t11-,12+,13+,15-;/m0./s1

InChI key

NBBQEIQEBNYSFK-PUILLJIJSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sophocarpine is a natural product found to have anti-inflammatory, anti-cancer, and antiviral activity. Sophocarpine was active in an mouse xenograft model. It inhibited growth of head and neck squamous cell carcinomas by downregulation of miR-21 expression. Sophocarpine was also shown to protecs mice from a form of hepatitis by inhibiting the IFNg/STAT1 pathway.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genhuan Yang et al.
Annals of palliative medicine, 9(4), 1622-1630 (2020-06-13)
Cigarette smoking is one of the main predisposing factors for atherosclerosis, which can lead to vascular inflammation. Emerging research has demonstrated that the rate of restenosis is increased following angioplasty in smokers. Sophocarpine, one of the Sophora alkaloids, can play

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service